Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 18 | 2528 | ||
NM_012470.3 | NP_036602.1 | 19 | 2720 | |||
NR_034053.2 | 20 | 2881 | ||||
XM_011515989.2 | XP_011514291.1 | 19 | 2322 | |||
GenBank mRNA | AF145029.1 | - | 19 | 2532 | ||
AJ133769.1 | - | 19 | 2317 | |||
AK001263.1 | - | 12 | 1274 | |||
AK022910.1 | - | 10 | 1054 | |||
AK225999.1 | - | 18 | 2499 | |||
AK312580.1 | - | 19 | 2447 | |||
BC009221.2 | - | 7 | 564 | |||
BC068530.1 | - | 19 | 2448 |