Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164114.1 | NP_001157586.1 | 6 | 2014 | ||
NM_001164115.1 | NP_001157587.1 | 5 | 865 | |||
NM_001164116.1 | NP_001157588.1 | 7 | 2200 | |||
NM_020356.3 | NP_065089.2 | 6 | 2176 | |||
XM_006723831.3 | XP_006723894.1 | 7 | 2600 | |||
GenBank mRNA | AJ276678.1 | - | 7 | 2197 | ||
AK295613.1 | - | 6 | 1879 | |||
AK308057.1 | - | 6 | 2177 | |||
AK314653.1 | - | 6 | 2109 |