Search
Search

Supt6| Gene Symbol: Supt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009297.2 | NP_033323.2 | 21 | 2812 | ||
| GenBank mRNA | AK129069.1 | - | 9 | 1135 | ||
| AK158846.1 | - | 2 | 596 | |||
| AK170533.1 | - | 21 | 2811 | |||
| BC072657.1 | - | 21 | 2732 | 30536338 | ||
| U40375.1 | - | 21 | 2810 | |||