Search
Search

Dtx3| Gene Symbol: Dtx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030714.2 | NP_109639.1 | 4 | 448 | ||
| GenBank mRNA | AB015425.1 | - | 3 | 430 | ||
| AK148444.1 | - | 4 | 467 | |||
| BC099687.1 | - | 4 | 448 | 6311735 | ||
| BC138304.1 | - | 3 | 327 | 8861323 | ||
| BC157899.1 | - | 3 | 329 | 9088091 | ||