Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003792.2 | NP_001003792.1 | 14 | 5074 | ||
NM_001003793.2 | NP_001003793.1 | 15 | 5128 | |||
NM_001177711.1 | NP_001171182.1 | 14 | 5080 | |||
XM_005265063.1 | XP_005265120.1 | 12 | 4785 | |||
XM_005265065.4 | XP_005265122.1 | 12 | 4478 | |||
XM_017006179.1 | XP_016861668.1 | 14 | 5529 | |||
XM_017006180.1 | XP_016861669.1 | 13 | 5481 | |||
XM_017006181.1 | XP_016861670.1 | 13 | 5478 | |||
XM_017006182.1 | XP_016861671.1 | 14 | 5557 | |||
GenBank mRNA | AK093863.1 | - | 1 | 69 |