Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321961.1 | NP_001308890.1 | 11 | 1621 | ||
NM_001321962.1 | NP_001308891.1 | 10 | 1435 | |||
NM_001321963.1 | NP_001308892.1 | 12 | 2106 | |||
NM_001321964.1 | NP_001308893.1 | 12 | 2431 | |||
NM_001321965.1 | NP_001308894.1 | 13 | 2744 | |||
NM_001321966.1 | NP_001308895.1 | 11 | 1739 | |||
NM_012472.5 | NP_036604.2 | 12 | 1681 | |||
NR_073525.2 | 13 | 1781 | ||||
NR_135905.1 | 12 | 1770 | ||||
NR_135906.1 | 9 | 1211 | ||||
NR_135907.1 | 11 | 1457 | ||||
NR_135908.1 | 8 | 1151 | ||||
NR_135909.1 | 11 | 1575 | ||||
NR_135910.1 | 11 | 1882 | ||||
NR_135911.1 | 9 | 1961 | ||||
NR_135912.1 | 12 | 2520 | ||||
NR_135913.1 | 11 | 2207 | ||||
XM_006716538.3 | XP_006716601.2 | 12 | 1645 | |||
XM_011516950.2 | XP_011515252.1 | 11 | 1585 | |||
XM_017013296.1 | XP_016868785.1 | 11 | 2314 | |||
XM_017013297.1 | XP_016868786.1 | 12 | 1799 | |||
XM_017013298.1 | XP_016868787.1 | 14 | 1726 | |||
GenBank mRNA | BG204280.1 | - | 2 | 408 | ||
DA570320.1 | - | 1 | 52 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U60666.1 | - | 2389 | Chr. - - [-] on Build |