Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN11 SIRNA Assays ›
          Gene Symbol:
          PTPN11
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 11
          Gene Aliases:
          BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
          Chromosome Location:
          Chr.12: 112418732 - 112509913 on Build GRCh38
          UniGene ID:
          Hs.506852
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11525
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002834.3 NP_002825.3 4 868
          NM_080601.1 NP_542168.1 4 868
          XM_006719526.2 XP_006719589.1 4 646
          XM_011538613.2 XP_011536915.1 4 682
          XM_017019722.1 XP_016875211.1 4 685
          GenBank mRNA AK289854.1 - 4 646
          BC008692.1 - 4 691 3872007
          BT007106.1 - 3 488
          D13540.1 - 4 644
          EU779706.1 - 1 155
          EU779708.1 - 3 482
          L03535.1 - 4 601
          L07527.1 - 4 641
          L08807.1 - 4 686
          X70766.1 - 4 617

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00275784_m1

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          activation of MAPK activity
          triglyceride metabolic process
          epidermal growth factor receptor signaling pathway
          integrin-mediated signaling pathway
          axonogenesis
          brain development
          heart development
          fibroblast growth factor receptor signaling pathway
          hormone-mediated signaling pathway
          regulation of phosphatidylinositol 3-kinase signaling
          cerebellar cortex formation
          platelet activation
          platelet formation
          T cell costimulation
          microvillus organization
          abortive mitotic cell cycle
          regulation of cell adhesion mediated by integrin
          negative regulation of cell adhesion mediated by integrin
          multicellular organism growth
          organ growth
          peptidyl-tyrosine dephosphorylation
          megakaryocyte development
          phosphatidylinositol-3-phosphate biosynthetic process
          atrioventricular canal development
          ERBB signaling pathway
          regulation of multicellular organism growth
          hormone metabolic process
          glucose homeostasis
          regulation of protein complex assembly
          positive regulation of mitotic cell cycle
          negative regulation of insulin secretion
          regulation of protein export from nucleus
          phosphatidylinositol phosphorylation
          positive regulation of hormone secretion
          platelet-derived growth factor receptor signaling pathway
          neurotrophin TRK receptor signaling pathway
          ephrin receptor signaling pathway
          phosphatidylinositol-mediated signaling
          multicellular organismal reproductive process
          genitalia development
          inner ear development
          homeostasis of number of cells within a tissue
          leukocyte migration
          negative regulation of cortisol secretion
          Bergmann glial cell differentiation
          negative regulation of growth hormone secretion
          face morphogenesis
          regulation of type I interferon-mediated signaling pathway
          intestinal epithelial cell migration
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of glucose import in response to insulin stimulus
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          non-membrane spanning protein tyrosine phosphatase activity
          SH3/SH2 adaptor activity
          insulin receptor binding
          protein binding
          1-phosphatidylinositol-3-kinase activity
          protein domain specific binding
          receptor tyrosine kinase binding
          D1 dopamine receptor binding
          phospholipase binding
          insulin receptor substrate binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          peptide hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0