Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00275784_m1
          See other PTPN11 GE Assays ›
          Gene Symbol:
          PTPN11
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 11
          Gene Aliases:
          BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
          Chromosome Location:
          Chr.12: 112418732 - 112509913 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00275784_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTPN11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002834.3 NP_002825.3 10 - 11 1611 83
          NM_080601.1 NP_542168.1 10 - 11 1611 83
          XM_017019722.1 XP_016875211.1 10 - 11 1428 83
          GenBank mRNA AK289854.1 - 10 - 11 1389 83
          BC008692.1 - 10 - 11 1434 3872007 83
          BT007106.1 - 9 - 10 1231 83
          D13540.1 - 10 - 11 1387 83
          L03535.1 - 10 - 11 1344 83
          L07527.1 - 10 - 11 1384 83
          L08807.1 - 10 - 11 1429 83
          S78088.1 - 1 - 2 127 83
          X70766.1 - 10 - 11 1360 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200008916.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000881.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Homo sapiens Human PDGF Pathway Human NGF Pathway HUMAN JAK-STAT PATHWAY HUMAN IL-6 PATHWAY Inventoried Human_PDGF_Pathway Human_NGF_Pathway Human_IL-6_Pathway Human_JAK-STAT_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PTPN11

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          activation of MAPK activity
          triglyceride metabolic process
          epidermal growth factor receptor signaling pathway
          integrin-mediated signaling pathway
          axonogenesis
          brain development
          heart development
          fibroblast growth factor receptor signaling pathway
          hormone-mediated signaling pathway
          regulation of phosphatidylinositol 3-kinase signaling
          cerebellar cortex formation
          platelet activation
          platelet formation
          T cell costimulation
          microvillus organization
          abortive mitotic cell cycle
          regulation of cell adhesion mediated by integrin
          negative regulation of cell adhesion mediated by integrin
          multicellular organism growth
          organ growth
          peptidyl-tyrosine dephosphorylation
          megakaryocyte development
          phosphatidylinositol-3-phosphate biosynthetic process
          atrioventricular canal development
          ERBB signaling pathway
          regulation of multicellular organism growth
          hormone metabolic process
          glucose homeostasis
          regulation of protein complex assembly
          positive regulation of mitotic cell cycle
          negative regulation of insulin secretion
          regulation of protein export from nucleus
          phosphatidylinositol phosphorylation
          positive regulation of hormone secretion
          platelet-derived growth factor receptor signaling pathway
          neurotrophin TRK receptor signaling pathway
          ephrin receptor signaling pathway
          phosphatidylinositol-mediated signaling
          multicellular organismal reproductive process
          genitalia development
          inner ear development
          homeostasis of number of cells within a tissue
          leukocyte migration
          negative regulation of cortisol secretion
          Bergmann glial cell differentiation
          negative regulation of growth hormone secretion
          face morphogenesis
          regulation of type I interferon-mediated signaling pathway
          intestinal epithelial cell migration
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of glucose import in response to insulin stimulus
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          non-membrane spanning protein tyrosine phosphatase activity
          SH3/SH2 adaptor activity
          insulin receptor binding
          protein binding
          1-phosphatidylinositol-3-kinase activity
          protein domain specific binding
          receptor tyrosine kinase binding
          D1 dopamine receptor binding
          phospholipase binding
          insulin receptor substrate binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          peptide hormone receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000351677.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000392597.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000530818.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000530818.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000530818.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000530818.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000531326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000531326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000531326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 ENST00000531326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_002834.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008916.hg.1 NM_080601.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0