Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hecw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108218.1 | NP_001101688.1 | 23, 24 | 4207 | ||
XM_006244912.2 | XP_006244974.1 | 23, 24 | 4392 |