Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000976.3 | NP_000967.1 | 6 | 530 | ||
GenBank mRNA | AF037643.1 | - | 6 | 502 | ||
AK026526.1 | - | 1 | 503 | |||
AK057602.1 | - | 4 | 2001 | |||
BC050644.1 | - | 1 | 483 | 5108312 | ||
BC059950.1 | - | 2 | 372 | 3454522 | ||
BC094831.1 | - | 1 | 478 | 5473344 | ||
BC105603.1 | - | 1 | 507 | 6698283 | ||
CB155565.1 | - | 6 | 530 | |||
L06505.1 | - | 6 | 483 |