Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLHC1 SIRNA Assays ›
Gene Symbol:
CLHC1
Gene Name:
clathrin heavy chain linker domain containing 1
Gene Aliases:
C2orf63
Chromosome Location:
Chr.2: 55172547 - 55232563 on Build GRCh38
UniGene ID:
Hs.347014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s43484
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLHC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135598.1
NP_001129070.1
8
1048
NM_152385.2
NP_689598.2
10
1499
XM_005264136.2
XP_005264193.1
9
1367
XM_006711937.3
XP_006712000.2
7
806
XM_006711938.3
XP_006712001.1
4
426
XM_006711939.3
XP_006712002.1
3
298
XM_011532519.2
XP_011530821.1
10
4392
XM_011532520.2
XP_011530822.1
10
1670
XM_011532521.2
XP_011530823.1
11
1826
XM_011532522.2
XP_011530824.1
11
1813
XM_011532523.2
XP_011530825.1
10
1902
XM_011532524.1
XP_011530826.1
9
1284
XM_011532525.2
XP_011530827.1
9
1350
XM_011532527.2
XP_011530829.1
10
4994
XM_011532528.1
XP_011530830.1
9
1894
XM_011532529.2
XP_011530831.1
8
1853
XM_011532530.1
XP_011530832.1
8
1614
XM_011532534.1
XP_011530836.1
8
1233
XM_011532535.2
XP_011530837.1
6
743
XM_017003333.1
XP_016858822.1
8
4194
XM_017003334.1
XP_016858823.1
8
1472
XM_017003335.1
XP_016858824.1
8
4562
XM_017003336.1
XP_016858825.1
9
1983
XM_017003337.1
XP_016858826.1
7
1072
XM_017003338.1
XP_016858827.1
7
1050
XM_017003340.1
XP_016858829.1
8
8494
XM_017003341.1
XP_016858830.1
8
1979
XM_017003342.1
XP_016858831.1
7
1480
XM_017003343.1
XP_016858832.1
7
1696
XM_017003344.1
XP_016858833.1
7
1784
XM_017003345.1
XP_016858834.1
6
1654
XM_017003347.1
XP_016858836.1
5
1522
XM_017003349.1
XP_016858838.1
5
3941
XM_017003350.1
XP_016858839.1
5
1452
GenBank mRNA
AK056000.1
-
3
445
AK315683.1
-
10
1457
BC029502.1
-
10
1499
5270720
BC036908.1
-
8
1048
5166064
BM046729.1
-
4
405
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0