Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006681.1 | NP_001006682.1 | 2 | 387 | ||
NM_001006682.1 | NP_001006683.1 | 2 | 335 | |||
NM_001006683.1 | NP_001006684.1 | 2 | 332 | |||
NM_001282461.1 | NP_001269390.1 | 2 | 397 | |||
NM_001282462.1 | NP_001269391.1 | 2 | 342 | |||
XM_005262008.4 | XP_005262065.2 | 2 | 181 | |||
XM_005262009.4 | XP_005262066.2 | 2 | 170 | |||
XM_005262010.1 | XP_005262067.1 | 2 | 392 | |||
XM_011530788.2 | XP_011529090.1 | 2 | 182 | |||
XM_011530789.2 | XP_011529091.1 | 2 | 203 | |||
XM_017029552.1 | XP_016885041.1 | 2 | 179 | |||
GenBank mRNA | AA319190.1 | - | 2 | 100 | ||
AF356353.1 | - | 2 | 204 | |||
BC000044.2 | - | 2 | 280 | 3504291 | ||
BC071974.1 | - | 2 | 319 | 6729986 | ||
BQ221120.1 | - | 344 | ||||
BX953266.1 | - | 2 | 316 |
Gene Symbol: SPIN2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019003.3 | NP_061876.3 | 2 | 447 | ||
XM_005262016.4 | XP_005262073.3 | 2 | 100 | |||
XM_005262017.4 | XP_005262074.3 | 2 | 100 | |||
XM_005262018.4 | XP_005262075.4 | 2 | 207 | |||
XM_005262019.3 | XP_005262076.1 | 2 | 1562 | |||
XM_017029598.1 | XP_016885087.1 | 2 | 1507 | |||
XM_017029599.1 | XP_016885088.1 | 2 | 221 | |||
GenBank mRNA | BC071694.1 | - | 2 | 447 | 5201217 | |
Y09858.1 | - | 1 | 491 |