Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013700.1 | NP_038728.1 | 12 | 1535 | ||
GenBank mRNA | AK154008.1 | - | 12 | 1524 | ||
BC066993.1 | - | 12 | 1513 | 5689229 |