Whole Allergen
i2 White-faced hornet
i2 White-faced hornet Scientific Information
Type:
Whole Allergen
Display Name:
White-faced hornet
Allergen code:
i2
Family:
Vespidae
Species:
Dolichovespula maculata
Route of Exposure:
Sting
Latin Name:
Dolichovespula maculata
Other Names:
Bald-faced hornet, Bald-faced aerial yellowjacket, Bald-faced wasp, Bald hornet, Blackjacket, White-tailed hornet, Spruce wasp, Bull wasp
WHO/ICD-11 code:
XE7TV
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
The white-faced hornet belongs to the genus Dolichovespula and is mainly found in the 48 contiguous states of the USA as well as Alaska, and throughout Canada. The white-faced hornet is actually a yellowjacket and not a true ‘hornet’. Dolichovespula species are amongst the most common culprits of severe immediate allergic reactions, along with honeybees, paper wasps, hornets, stinging ants and bumblebees.
Allergen
Nature
The white-faced hornet belongs to the genus Dolichovespula and is mainly found in the 48 contiguous states of the USA as well as Alaska, and throughout Canada. The white-faced hornet is actually a yellowjacket and not a true ‘hornet’ (1). It builds aerial nests, which are usually found in exposed places such as hanging from tree branches. These yellowjackets usually feed on live prey, particularly spiders and flies (2).
Taxonomy 
Taxonomic tree of White Faced Hornet (3)
Domain
Eukaryota
Kingdom
Animalia
Phylum
Arthropoda
Subphylum
Hexapoda
Class
Insecta
Order
Hymenoptera
Family
Vespidae
Genus
Dolichovespula Rohwer
Epidemiology
Worldwide distribution 
The white-faced hornet is the largest and most widespread of the North American Vespidae (4) and are amongst the most common culprits of severe immediate allergic reactions, along with honeybees, paper wasps, hornets, stinging ants and bumblebees (5).
Clinical Relevance
Dolichovespula maculata belongs to the order Hymenoptera, one of the most frequent triggers for severe IgE-mediated anaphylaxis in adults. Its sting is not only very painful, but may also induce severe allergic responses in humans (4). Systemic reactions may be restricted to generalized symptoms of the skin, but can also affect the respiratory and vascular system and lead to multi-organ failure (6). 
Molecular Aspects
Allergenic molecules
Table adapted from Allergome.org (7).
Allergen
Type
Mass (kDa)
Dol m 1
Phospholipase A1
38
Dol m 2
Hyaluronidase
41
Dol m 5
Unknown
23
The allergens of the white-faced hornet include phospholipase A1 (Dol m I) and antigen 5 (originally Dol m V, now Dol m 5) (8). Antigen 5 (Dol m 5) is a protein of approximately 23 kDa first identified by King et al, in 1978. Antigen 5 is frequently reported to be the most allergenic venom component in different species of social wasps, including those belonging to the genus Dolichovespula, Vespa, Vespula, Polistes and Polybia (9). However, the role of antigen 5 is unknown (6). Other identified allergens include hyaluronidase (Dol m 2, 43 kDa) and phospholipase A1 (Dol m 1, 37 kDa) (8).
Cross-reactivity
Antigen 5 allergens of different Vespinae members (including Dolichovespula, Vespula and Vespa species) display pronounced cross-reactivity (9).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr.Michael Thorpe
Last reviewed: May 2022
References
  1. PennState. Baldfaced Hornet 2021 [cited 2021 3.12.21]. Available from: https://extension.psu.edu/baldfaced-hornet.
  2. Kimsey LS, Carpenter JM. The Vespinae of North America (Vespidae, Hymenoptera). Journal of Hymenoptera Research. 2012;28:37-65.
  3. ITIS. Dolichovespula maculata  (Linnaeus) 2021 [cited 2021 30.11.21]. Available from: https://www.itis.gov/servlet/SingleRpt/SingleRpt?search_topic=TSN&search_value=154260#null.
  4. Schmidt JO, Blum MS. Toxicity of Dolichovespula maculata venom. Toxicon. 1979;17(6):645-8.
  5. Korošec P, Jakob T, Harb H, Heddle R, Karabus S, de Lima Zollner R, et al. Worldwide perspectives on venom allergy. The World Allergy Organization journal. 2019;12(10):100067-.
  6. Blank S, Bazon ML, Grosch J, Schmidt-Weber CB, Brochetto-Braga MR, Bilò MB, et al. Antigen 5 Allergens of Hymenoptera Venoms and Their Role in Diagnosis and Therapy of Venom Allergy. Curr Allergy Asthma Rep. 2020;20(10):58.
  7. Allergome. Dolichovespula maculata 2021 [cited 2021 30.11.21]. Available from: https://www.allergome.org/script/search_step2.php.
  8. Soldatova L, Kochoumian L, King TP. Sequence similarity of a hornet (D. maculata) venom allergen phospholipase A1 with mammalian lipases. FEBS Lett. 1993;320(2):145-9.
  9. Bazon ML, Silveira LH, Simioni PU, Brochetto-Braga MR. Current Advances in Immunological Studies on the Vespidae Venom Antigen 5: Therapeutic and Prophylaxis to Hypersensitivity Responses. Toxins. 2018;10(8).