Type:
Whole Allergen
Whole Allergen
Whole Allergen
White-faced hornet
i2
Vespidae
Dolichovespula maculata
Sting
Dolichovespula maculata
Bald-faced hornet, Bald-faced aerial yellowjacket, Bald-faced wasp, Bald hornet, Blackjacket, White-tailed hornet, Spruce wasp, Bull wasp
XE7TV
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
The white-faced hornet belongs to the genus Dolichovespula and is mainly found in the 48 contiguous states of the USA as well as Alaska, and throughout Canada. The white-faced hornet is actually a yellowjacket and not a true ‘hornet’. Dolichovespula species are amongst the most common culprits of severe immediate allergic reactions, along with honeybees, paper wasps, hornets, stinging ants and bumblebees.
The white-faced hornet belongs to the genus Dolichovespula and is mainly found in the 48 contiguous states of the USA as well as Alaska, and throughout Canada. The white-faced hornet is actually a yellowjacket and not a true ‘hornet’ (1). It builds aerial nests, which are usually found in exposed places such as hanging from tree branches. These yellowjackets usually feed on live prey, particularly spiders and flies (2).
Taxonomic tree of White Faced Hornet (3) |
|
Domain |
Eukaryota |
Kingdom |
Animalia |
Phylum |
Arthropoda |
Subphylum |
Hexapoda |
Class |
Insecta |
Order |
Hymenoptera |
Family |
Vespidae |
Genus |
Dolichovespula Rohwer |
The white-faced hornet is the largest and most widespread of the North American Vespidae (4) and are amongst the most common culprits of severe immediate allergic reactions, along with honeybees, paper wasps, hornets, stinging ants and bumblebees (5).
Dolichovespula maculata belongs to the order Hymenoptera, one of the most frequent triggers for severe IgE-mediated anaphylaxis in adults. Its sting is not only very painful, but may also induce severe allergic responses in humans (4). Systemic reactions may be restricted to generalized symptoms of the skin, but can also affect the respiratory and vascular system and lead to multi-organ failure (6).
Table adapted from Allergome.org (7).
Allergen |
Type |
Mass (kDa) |
Dol m 1 |
Phospholipase A1 |
38 |
Dol m 2 |
Hyaluronidase |
41 |
Dol m 5 |
Unknown |
23 |
The allergens of the white-faced hornet include phospholipase A1 (Dol m I) and antigen 5 (originally Dol m V, now Dol m 5) (8). Antigen 5 (Dol m 5) is a protein of approximately 23 kDa first identified by King et al, in 1978. Antigen 5 is frequently reported to be the most allergenic venom component in different species of social wasps, including those belonging to the genus Dolichovespula, Vespa, Vespula, Polistes and Polybia (9). However, the role of antigen 5 is unknown (6). Other identified allergens include hyaluronidase (Dol m 2, 43 kDa) and phospholipase A1 (Dol m 1, 37 kDa) (8).
Antigen 5 allergens of different Vespinae members (including Dolichovespula, Vespula and Vespa species) display pronounced cross-reactivity (9).
Author: RubyDuke Communications
Reviewer: Dr.Michael Thorpe
Last reviewed: May 2022