Type:
Whole Allergen
Whole Allergen
Whole Allergen
Ethylene oxide
k78
Ethylene oxide
XM4G15
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Ethylene oxide is commonly used industrially for sterilization of heat sensitive medical devices.
Unexpected exposure
Patients undergoing chronic haemodialysis and those exposed to tubings, syringes and other material sterilized with ethylene oxide.
Environment exposure
Any workplace where ethylene oxide is used in significant amounts. Certain medical wards
Severe allergic symptoms rhinitis, asthma, chest oppression, and urticaria and even anaphylaxis triggered by IgE antibodies to ethylene oxide (1-2) have been reported (3-4). IgE-mediated anaphylaxis mediated by ethylene oxide has been published in patients regularly undergoing haemodialysis and approximately 10% of such patients have been found to have IgE antibodies to ethylene oxide (5-6)
Last Reviewed- April 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.