Whole Allergen
o203 TetraMin®, Fish feed
o203 TetraMin®, Fish feed Scientific Information
Type:
Whole Allergen
Display Name:
TetraMin®, Fish feed
Allergen code:
o203
Family:
Various
Summary
Occupational allergen
An occupational allergen, which may result in allergy symptoms in sensitised individuals.
Exposure
Allergen Exposure
Geographical distribution
TetraMin® is a leading fish food made from 40 ingredients and marketed as a “complete feed for hobby pet fish”. It contains the following components: molluscs and crustaceans, fish and fish by-products, grain, yeasts, vegetable protein extracts, oils, fats, seaweed, sugar, lecithin, and permissible foodstuff colourings and antioxidants. The manufacturers describe the product as comprising protein (46%), fat (8%), fibre (2%), ash (11%), and water (6%).
Allergens
No allergens have yet been characterised.
Clinical Experience
IgE-mediated reactions
TetraMin® may uncommonly induce symptoms of skin or inhalant allergy in sensitised individuals. No reports of hypersensitivity reactions to Tetramin® have been made in the peer-reviewed medical literature; however, as a consequence of its ingredients, e.g., crustaceans, which are important allergens, these reactions are possible.