Type:
Whole Allergen
Whole Allergen
Whole Allergen
TetraMin®, Fish feed
o203
Various
An occupational allergen, which may result in allergy symptoms in sensitised individuals.
TetraMin® is a leading fish food made from 40 ingredients and marketed as a “complete feed for hobby pet fish”. It contains the following components: molluscs and crustaceans, fish and fish by-products, grain, yeasts, vegetable protein extracts, oils, fats, seaweed, sugar, lecithin, and permissible foodstuff colourings and antioxidants. The manufacturers describe the product as comprising protein (46%), fat (8%), fibre (2%), ash (11%), and water (6%).
No allergens have yet been characterised.
TetraMin® may uncommonly induce symptoms of skin or inhalant allergy in sensitised individuals. No reports of hypersensitivity reactions to Tetramin® have been made in the peer-reviewed medical literature; however, as a consequence of its ingredients, e.g., crustaceans, which are important allergens, these reactions are possible.
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Contact us
Interested in utilizing our solutions in your laboratory or healthcare practice?
Not a lab professional or healthcare provider but would like to learn more about allergy testing?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.