Case Study
Post-op: PCT Decline Reflects Host Response to Antibiotics 
Critical Care Medicine, Surgery
Learn more about implementing procalcitonin testing
in your hospital.
Explore similar topics
Key Takeaways
Takeaway #1
Takeaway #1
PCT levels are elevated post-op for 24 to 36 hours, then quickly trend down to normal.1
Takeaway #2
Takeaway #2
A PCT result of 160 μg/L was extremely elevated, indicative of bacterial burden (> 5 ng/mL).2 The elevated PCT finding of 160 μg/L can aid in the differential diagnosis: Hypovolemia vs. peritonitis. The patient was taken back to the OR whereby bowel perforation secondary to anastomotic leak was confirmed and repaired.
Takeaway #3
Takeaway #3
Post-op, antibiotics were initiated Vancomycin, Imipenem, and Moxifloxacin, serial PCT levels were monitored and trended down, of note there was a continual decrease of > 50% daily confirming the patient was responding to therapy.
Share your Clinical Experience
Submit your own PCT Case Study and you may be selected to be featured on procalcitonin.com. 
Submit a case study here
Share your experience
Related Resources
References
  1. B·R·A·H·M·S GmbH (part of Thermo Fisher Scientific), B·R·A·H·M·S PCT sensitive KRYPTOR® Instruction for Use (Version 19.0us) [Internet]. 2018. Available from: https://assets.thermofisher.com/TFS-Assets/CDD/Package-Inserts/IFU-825-PCT-sensitive-KRYPTOR-HN-CUS-0888-R19-EN-US-USA-19-EN-US.pdf.
  2. Giaccaglia V, Salvi PF, Cunsolo GV, Sparagna A, Antonelli MS, Nigri G, et al. Procalcitonin, as an early biomarker of colorectal anastomotic leak, facilitates enhanced recovery after surgery. J Crit Care. 2014 Aug;29(4):528-32.