Gibco high-quality selection agents offer unique solutions for your research needs, such as dual selection and rapid stable cell line establishment.
- View our full list of antibiotics and antimycotics
Which selection antibiotic is right for you?
Eukaryotic selection antibiotics
Selection antibiotic | Most common selection usage | Common working concentration | Available powder sizes | Available liquid sizes | Order |
---|---|---|---|---|---|
Blasticidin | Eukaryotic and Bacteria | 1-20 µg⁄ml | 50 mg | 10x1 ml, 20 ml | Order |
Geneticin (G-418) | Eukaryotic | 100 - 200 µg ⁄ ml bacteria 200 – 500 µg ⁄ ml mammalian cells | 1 g, 5 g, 10 g, 25 g | 20ml, 100ml | Order |
Hygromycin B | Dual-selection Experiments and Eukaryotic | 200-500 µg ⁄ ml | - | 20ml | Order |
Mycophenolic Acid | Mammalian and Bacteria | 25 µg ⁄ ml | 500 mg | - | Order |
Puromycin | Eukaryotic and Bacteria | 0.2 – 5 µg ⁄ ml | - | 10 x 1 ml, 20ml | Order |
Zeocin | Mammalian, insect, yeast, bacteria and plants | 50 – 400 µg ⁄ ml | - | 8 x 1.25 ml, 50 ml | Order |
Bacterial selection antibiotics
Selection antibiotic | Most common selection usage | Common working concentration | Available powder sizes | Available liquid sizes | Order |
---|---|---|---|---|---|
Actinomycin D | Bacteria | 1 µg ⁄ ml | 5 mg, 10 mg | - | Order |
Ampicillin Sodium Salt | Bacteria | 10 – 25 µg ⁄ ml | 200 mg | - | Order |
Blasticidin | Eukaryotic and Bacteria | 50-100 µg ⁄ ml | 50 mg | 10x1ml, 20ml | Order |
Carbenicillin, Disodium Salt | Agrobacterium and E. coli | 100 – 500 µg ⁄ ml | 5 g | - | Order |
Kanamycin Sulfate | Bacteria | 100 µg ⁄ ml | 5 g, 25 g | 100ml | Order |
Mycophenolic Acid | Mammalian and Bacteria | 25 µg ⁄ ml | 500 mg | - | Order |
Neomycin Sulfate | Bacteria | 50 µg ⁄ ml | 100 g | - | Order |
Polymyxin B Sulfate | Bacteria | 100 units / ml | 25 MU | - | Order |
Puromycin | Eukaryotic and Bacteria | 0.2 – 5 µg ⁄ ml | - | 10 x 1 ml, 20ml | Order |
Streptomycin Sulfate | Bacteria | 50 – 100 µg ⁄ ml | - | 100 g | Order |
Zeocin | Mammalian, insect, yeast, bacteria and plants | 75 – 400 µg ⁄ ml | - | 8 x 1.25 ml, 50 ml | Order |