Search
Learn the fundamentals of protein expression using the Gibco ExpiSf Expression System with detailed overviews of the unique system components and key workflow steps for which optimal techniques will help maximize results. This protocol guide includes required material checklists, visual step-by-step guides, and helpful tips.
At the end of this protocol guide, you should be able to:
The ExpiSf Expression System is the first chemically defined baculovirus expression system for high-yielding protein expression in Sf9 insect cells. Undefined media components like hydrolysates can make it quite difficult to get consistent results with Sf9 cells. The ExpiSf Expression System has been optimized to work with chemically defined Gibco ExpiSf CD Medium, which is free of hydrolysates, serum, protein, and animal-origin components. Transfection, baculovirus production, and recombinant protein expression all take place directly in ExpiSf CD Medium, providing consistency from start to finish.
ExpiSf Expression System Starter Kits provide enough cells, culture medium, and reagents to infect 1 L of cell culture. The kits also include the Gibco Bac-to-Bac Baculovirus Expression System for generation of bacmid DNA. With the exception of Gibco MAX Efficiency DH10Bac Competent Cells, all components of ExpiSf Expression System Starter Kits are animal origin–free (AOF).
Component |
Quantity |
Storage |
ExpiSf9 Cells (1 x 107 cells/mL) |
2 x 1.5 mL |
Liquid nitrogen, vapor phase* |
1 L |
2°C to 8°C (protect from light) |
|
ExpiSf Enhancer |
4 x 1 mL |
2°C to 8°C (protect from light) |
1 mL |
2°C to 8°C (do not freeze) |
|
500 mL |
2°C to 8°C (protect from light) |
|
pFastBac 1 Expression Vector (0.5 µg/µL)** |
10 µg |
–20°C |
pFastBac 1-Gus Control Vector (0.2 ng/µL)** |
4 ng |
–20°C |
5 x 100 µL |
Competent cells: –80°C pUC19 DNA: –20°C SOC Medium: 4°C or room temperature |
|
Nalgene Single-Use PETG 125 mL Erlenmeyer Flasks (vented, non-baffled) |
24 |
Room temperature |
* Store the frozen cells in liquid nitrogen until ready to use. Do not store the cells at −80°C.
** In TE buffer, pH 8.0: 10 mM Tris-HCl, 1 mM EDTA, pH 8.0.
Recommended consumables, instruments, and equipment for protein expression with the ExpiSf Expression System.
Key laboratory instruments and equipment |
Key laboratory consumables |
Biosafety cabinet with HEPA filter Incubator with temperature, humidity, and CO2 regulators Reagents and equipment for determining viable cell density and percent viability (automated cell counter, trypan blue solution) Aspiration pump (peristaltic or vacuum) Waste containers and vacuum traps |
Cell culture flasks, vented, non-baffled (125 mL to 5 L) Serological pipettes and pipettors (1 mL to 50 mL) Multichannel pipettors and pipettes (1 µL to 200 µL) Single channel pipettors and pipettes (1 µL to 1 mL) Cell culture medium, PBS, and sterile H2O Personal protective equipment (PPE) (laboratory coat, safety goggles, disposable gloves, eye wash station) |
General cell handling |
|
|
|
|
|
|
|
|
|
|
|
|
|
Note: Do not subculture cells until they reach a density of at least 5 x 106 viable cells per mL. Cells that are subcultured at lower densities can have longer doubling times and produce lower titers over time. Modify the seeding density to attain the target cell density of at least 5 x 106 viable cells per mL at the subculturing stage. |
|
|
|
Medium |
Important: ExpiSf CD Medium is light-sensitive. For optimal results, protect the medium from light during use and storage. |
Thaw cells (Day 1)
|
1. Remove the vial of cells from liquid nitrogen and swirl it in a 37°C water bath for 1–2 minutes to thaw the cells until only a small amount of ice remains. Do not submerge the vial in the water bath. 2. Just before the cells are completely thawed, decontaminate the exterior surface of the vial by wiping it down with 70% ethanol. When thawing is complete, open the vial in a laminar flow hood. |
Add cells to medium (Day 1)
|
3. Use a 2 mL or 5 mL pipette to transfer the entire contents of the vial into the sterile 125 mL Erlenmeyer shake flask containing 25 mL of ExpiSf CD Medium warmed to room temperature. |
Incubate cells
|
4. Incubate the cells on the orbital shaker at 27°C in the non-humidified, air-regulated incubator. Note: Set the shaking speed to 125 ± 5 rpm for shakers with a 19 mm or 25 mm shaking diameter or 95 ± 5 rpm for shakers with a 50 mm shaking diameter. |
Count cells and determine viability (Day 3–5)
|
5. Three days post-thaw, determine the viable cell density and percent viability. Cell viability should be ≥80% by Day 3 post-thaw. 6. Continue to monitor cell density and viability. Subculture the cells once the culture has reached 5 x 106 to 10 x 106 viable cells per mL, typically 4–5 days post-thaw. |
Table 1. Recommended seeding densities for routine maintenance of ExpiSf9 Cells.
Subculture timing |
Recommended seeding density |
For cells ready 3 days post-subculture |
0.7 x 106 to 1.0 x 106 viable cells per mL |
For cells ready 4 days post-subculture |
0.4 x 106 to 0.6 x 106 viable cells per mL |
Table 2. Recommended volumes and shaking speeds for routine maintenance of ExpiSf9 Cells.
Flask size |
Culture volume |
Shaking speed* |
125 mL |
25–30 mL |
19 mm shaking diameter: 125 ± 5 rpm 25 mm shaking diameter: 125 ± 5 rpm 50 mm shaking diameter: 95 ± 5 rpm |
250 mL |
50–60 mL |
|
500 mL |
100–120 mL |
|
1 L |
200–240 mL |
|
2 L |
400–480 mL |
|
3 L |
600–800 mL |
80 ± 5 rpm (19 mm shaking diameter) |
* Use upper ranges of recommended shaking speeds for larger culture volumes.
Passage cells (Day 1)
|
1. Use the viable cell density to calculate the volume of cell suspension required to seed a new shake flask according to the recommended seeding densities in Table 1 and the recommended culture volumes in Table 2. 2. Transfer the calculated volume of cells to fresh ExpiSf CD Medium warmed to room temperature in a shake flask. |
Subculture cells
|
3. Incubate the cells on an orbital shaker in the 27°C non-humidified incubator until they reach a density of 5 x 106 to 10 x 106 viable cells per mL. Note: Cells that are subcultured at densities outside this log phase growth window may have longer doubling times and produce lower protein titers over time. Modify the initial seeding density to attain the target cell density of 5 x 106 to 10 x 106 viable cells per mL at the subculturing stage. 4. Repeat Steps 1–3 to maintain or expand the cells for transfection or infection. |
|
Note: If you want to freeze ExpiSf9 Cells, harvest the cells when they reach a viable cell density of 3 x 106 to 4.5 x 106 viable cells per mL with ≥90% viability. This will typically be on Day 3 after passaging cells from a seeding density of 0.5 x 106 to 0.6 x 106 cells/mL. If the viable cell density is too low at the time of harvest, return the cells to the incubator for an extra day. If the viable cell density is too high at the time of harvest, subculture the cells at 0.5 x 106 to 0.6 x 106 cells/mL and prepare for harvest again after 3–4 days. |
For Research Use Only. Not for use in diagnostic procedures.