Cryoware-230x195
Streamline your cryogenic storage
Thermo Scientific™ Cryoboxes protect and organize your important samples and are available in materials ranging from economical cardboard to break-resistant, durable polycarbonate to meet any need and budget. The Dense Storage format is available to maximize storage, holding 169 1.0mL Thermo Scientific™ Nunc™ Cryobank™ tubes. All Thermo Scientific Cryoboxes are designed to provide dependable sample protection and meet the rigorous requirements of cryogenic storage.
Please begin by selecting a filter below to see recommended products.
Filter Results
Recommended product(s)
Support
Need technical support? Contact our expert team for technical and application support of laboratory products.