榆樹 過敏原基本資料、症狀和治療
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
榆樹屬榆科,包括大約 35 種林木和觀賞性遮蔭樹。榆樹原產於北方溫帶地區,透過風媒傳粉。1,2 榆樹是過敏的主要來源,一年之中幾乎任何時候都可以授粉,具體取決於樹種和地點。3
榆樹通常分佈於北美和歐亞大陸溫帶地區、美洲和非洲以及東南亞熱帶地區。5
許多榆樹過敏患者在接觸其他過敏原 (如木本、雜草或草本花粉) 時也會出現症狀,因此很難確定是哪種花粉引起症狀,尤其是花粉季節有重疊時。這稱為交叉反應,當人體的免疫系統認定不同物質中的蛋白質或成分在結構上相似或生物學上相關,就會發生交叉反應,從而引發免疫反應。9 其他可能導致榆樹花粉過敏的呼吸道過敏原包含梣葉槭樹和其他特定樹木、雜草和草本花粉。10
*在您的國家/地區,這些產品可能並未獲准用於臨床用途。請與醫護人員溝通,以了解其可用性。
過敏性鼻炎的控制方法包括避開相關過敏原、對症治療和過敏原免疫療法。6-8
榆樹過敏症狀可能與許多其他花粉過敏相似,可能包括:4
如果您對榆樹過敏並患有氣喘,則木本花粉可能會觸發或加劇氣喘症狀,例如咳嗽和喘鳴。4
結合您的症狀史,皮膚點刺檢測或血清特異性 IgE 檢測有助於確定您是否對特定過敏原敏感。如果您確診患有過敏症,醫護人員將與您一起制定控制計畫。
*在您的國家/地區,這些產品可能並未獲准用於臨床用途。請與醫護人員溝通,以了解其可用性。
根據樹種,榆樹可在兩個不同時間開花和產生花粉。大多數樹種在南緯地區為 1 月或 2 月進行授粉,而在北緯地區則遲至 4 月。然而,南部地區的一些榆樹在夏末甚至 11 月才開花。這些秋季授粉的樹種比春季授粉的樹種更容易引起過敏。3