Search
Engineered to facilitate accurate genotypic microbial identification in less than five hours, MicroSEQ PCR and Sequencing Reagent Kits help provide a streamlined, high-resolution workflow from DNA amplification through species-level identification. Designed for environmental monitoring and regulated quality control laboratories, these microbial identification kits are built to facilitate answers without unnecessary uncertainty, giving teams the clarity and confidence on demand they need when it matters most.
Each kit includes proprietary master mixes, optimized primers, and all required reagents to simplify implementation and support consistent PCR and sequencing performance. Validated for use with the SeqStudio family of genetic analyzers, MicroSEQ kits enable high-resolution Sanger sequencing across a broad range of bacterial and fungal organisms. Engineered to limit ambiguity, reducing repeat testing, and supporting root cause identification on demand, the MicroSEQ system helps laboratories improve predictability, scale operations efficiently, and reduce operational risk over time. The result is reliable, reproducible species-level identification that strengthens contamination investigations and supports confident regulatory decision-making.
MicroSEQ kits include:
PCR Kits |
Sequencing Kits |
|||
2X Master Mix |
Positive control DNA |
Negative control DNA |
3X Forward sequencing mix |
3X Reverse sequencing mix |
|
|
|
|
|
For bacterial identification, the MicroSEQ system targets the 16S ribosomal RNA gene, a universally conserved genetic marker present in all bacteria. By combining conserved regions for reliable amplification with hypervariable regions for differentiation, this approach enables accurate, species-level identification across a broad range of organisms encountered in environmental monitoring and quality control laboratories.
To support both routine workflows and complex investigations, the MicroSEQ system provides flexible sequencing options:
By enabling both routine and high-resolution identification within the same standardized workflow, the MicroSEQ system allows laboratories to scale their approach based on the needs of each sample without changing platforms or processes.
The result is clear, species-level bacterial identification that supports confident decision-making, improves investigation efficiency, and helps laboratories avoid the delays and uncertainty associated with incomplete or low-confidence results.
Figure 1. Hypervariable regions within the 16S rRNA gene. There are 9 hypervariable regions within the bacterial 16S gene, indicated in green. The conserved regions are indicated in grey.
Description |
PCR kit |
Sequencing kit |
Library |
# of entries |
16sR full gene |
MicroSEQ ID 16S rDNA Full Gene Library v2.1 |
1,262 |
||
16sR, first 500 base pairs |
MicroSEQ ID 16S rDNA 500 Microbial Library, v2022 |
2,100 |
||
MicroSEQ ID 16S rDNA 500 Supplemental Library, v2022 |
7,645 |
Fungal identification continues to evolve as environmental monitoring programs encounter a broader and more diverse range of molds, yeasts, and closely related species. Increasing organism complexity, combined with growing regulatory expectations for species-level identification, makes accurate fungal classification more challenging than ever.
These challenges often lead to ambiguous identifications, multiple matches, or low-confidence results, resulting in repeat testing, extended investigations, and delays in root cause determination.
To address this, the MicroSEQ system facilitates flexible fungal identification options within a single workflow:
By enabling LSU-D2, ITS, or both within the same standardized workflow, the MicroSEQ system allows laboratories to match the level of resolution to the complexity of the organism without changing platforms or processes.
The result is more reliable fungal identification, improved investigation timelines, and clear, defensible results that support contamination control, regulatory compliance, and confident decision-making in high-stakes environments.
| MicroSEQ D2 rDNA Fungal Sequencing Kit | MicroSEQ ITS rDNA Fungal Identification Kit |
| Fungal identification based on 'legacy' D2 region | Fungal identification based on 'hyper variable' ITS region |
| Reliable target for routine fungal and yeast identification | Enhanced resolution for closely related molds |
| Proven performance in QC workflows | Greater sequence variability for improved species discrimination |
| Effective for many common fungal isolates | Designed for higher specificity and accuracy |
| Supported by validated MicroSEQ D2 library of 1,737 sequences | Supported by 38,000+ curated ITS sequences |
| Ideal for standardized routine testing | Ideal when higher discriminatory power is required |
| Integrated in MicroSEQ microbial identification workflow | |
Description |
PCR kit |
Sequencing kit |
Library |
# of entries |
LSU-D2 rDNA |
MicroSEQ ID Fungal Gene Library, v3.0 |
1,735 |
||
| ITS rDNA | MicroSEQ ITS rDNA Fungal Identification Kit | MicroSEQ ITS rDNA Fungal Identification Kit + Prepman Ultra reagent | MicroSEQ ID Fungal ITS Gene Library, v1.0 | 38,229 |
No matter your questions, we are here to help. Our experienced team of scientists and technical specialists is ready to assist you in choosing the right solution and products. Complete this form to start the conversation.
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.
For Research Use Only. Not for use in diagnostic procedures.