Leverage a dedicated analyzer for Special Protein Assays
Leverage a dedicated analyzer for Special Protein Assays
Increase productivity and efficiency for both clinical chemistry and special protein workflows, with the value brought by the Optilite® Analyser
A dedicated protein analyzer is complementary to a clinical chemistry analyzer
Clinical diagnostic labs are under increasing pressure facing a range of challenges. From rising workloads to increased administrative and regulatory burden, all while dealing with the shortage of skilled staff.
With the combination of these challenges, laboratory managers could decide to consolidate all chemistry and immunoassays onto a single clinical chemistry platform. While this offers benefits such as automating pre-analytical steps, combined training, and maximizing laboratory space, using a clinical chemistry analyzer isn’t always the best approach for special protein testing.
Benefits of using a dedicated analyzer for special protein assays
Special protein tests often come with more complex assay protocol requirements, including:
  • Wide measuring ranges
  • Multiple dilutions
  • Automated antigen excess detection
  • Suited for low-volume samples such as CSF
Running these assays on clinical chemistry analyzers causes extra steps and manual interventions leading to:
  • Disruption of desired automated workflows
  • Need for offline manual dilutions
  • Additional wash cycles to prevent cuvette fogging
  • Higher frequency of maintenance
  • Longer turnaround times for clinical chemistry assays waiting for longer protocols to finish
Thumbnail of the Hennepin case study front page
Download the Hennepin case study
In this case study, Hennepin County Medical Center measured the benefits of moving special protein testing from the clinical chemistry system to the dedicated Optilite Analyser.
Performing special protein diagnostics on a dedicated protein analyzer results in:
Optimized workflows delivering optimal turnaround times for both special protein and clinical chemistry testing
Cost efficiency through optimized dilution steps, reagent stability and packaging
Minimal hands-on time, user interventions and training, reducing the burden on staff
Increased throughput from optimized protocols, intelligent sample scheduling and continuous access
Standalone instrument requiring limited laboratory space (< 0.7m2)
Ability to offer a more complete menu in special protein testing
Why choose Optilite as a Special Protein Analyzer?
Due to the nature of special protein assays and the specific protocols they require, the most optimal setting is to perform these assays on a dedicated platform. Why is that the case? What are the benefits of implementing a standalone protein analyzer?
Increased productivity
Eliminating the need for manual, offline dilutions leads to shorter turnaround times and by consequence a higher throughput. Depending on the assay and the concentration of the samples, the turnaround time can be improved by as much as 82%.1,2 Avoiding the need for special washing and minimizing the impact of cuvette fogging will avoid any unnecessary downtime of the routine clinical chemistry system.1
Better workflow efficiency
The benefits of not requiring offline dilutions are evident and contribute to a more streamlined and automated workflow for special protein assays. Another important gain lies in optimized protocols where wider measuring ranges and optimized dilution steps reduce the number of tests per result and deliver a saving on reagent costs.1,2
Quality and breadth of your protein testing service
Having a dedicated protein analyzer allows you to run a broader panel of assays in your own lab, reducing the need, time and cost associated with sending out certain tests.
The optimized protocols allow for more automated antigen excess detection increasing your confidence to obtain a first-time right result.
A dedicated menu offering from a specialized partner means the packaging and open vial stability is developed to fit with the volumes and testing frequency of these special protein assays.
All of these benefits can be achieved with the Optilite Analyser and its optimized, high-quality assay menu
Read the Omosule case study
Customer stories
Hear more about the value the Optilite Analyser has brought to these customers in the videos below:
Saint Antonius Hospital, Netherlands
Listen to why Mr. Hans De Graaf considers Binding Site to be the market leader in the field of special protein diagnostics.
Sonic Healthcare, Belgium
Hear Dr. Astrid Coppens explain the value that the Optilite Analyser brought to their high volume laboratory.
Istituto Diagnostico Varelli, Italy
Learn the benefits of working with the Optilite Analyser and how Dr. Marco Varelli values the support and collaboration from Binding Site.
What our customers say
With over 1700 instruments installed worldwide, many customers are reaping the range of benefits delivered by the Optilite Analyser
Get in touch with us if you are interested in discussing the workflow benefits for your lab
Technical & scientific expertise
Our technical and scientific expertise spans from R&D, all the way to production.
98 %
of respondents give maximum score for service responsiveness³
99 %
of respondents give maximum score for level of service³
97 %
Quality rating of end-user testing⁴
Learn all you need to know about Optilite Analyser
Fully optimized for simplified, high-quality Special Protein Analysis.
Contact Us?
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you
Optilite is a registered trademark of The Binding Site Group Ltd (Birmingham, UK) in certain countries.
Other brand or product names may be trademarks of their respective holders.
References
  1. Saenger et al. Improvement in Workflow and Efficiency for Measurement of Special Proteins Using the Optilite® Standalone Automated Analyzer. Performed at Hennepin County Medical Center, Minneapolis, USA. Presented at the 2021 AACC Annual Scientific Meeting & Clinical Lab Expo, September 26-30, 2021 in Atlanta, Georgia
  2. Omosule et al. Method Comparison and Workflow Differences Using the Same Free Light Chain Assay on 2 Analyzer Platforms, The Journal of Applied Laboratory Medicine, 2023.
  3. Based on end user feedback following service repairs and applications visits between October 2022 and September 2023
  4. Based on feedback from 20 end user following training given between April – June 2023