Transform your diagnostic testing with the Optilite® Analyser
The robust and proven analyzer from The Binding Site, bringing you a reliable and optimized solution for special protein testing
Load.
Start.
Results.
Simplified workflow for special protein testing
Specialized
Unique and extensive assay menu developed and supported by dedicated local and global teams of technical and scientific experts
Quality
Fully automated dilutions eliminate a source of user error, and optimized antigen excess protection can prevent reporting of falsely low or even false negative results
Productivity
Efficient handling of high sample workload and minimal operator intervention frees up valuable lab staff time
Cost efficiency
Optimized measuring ranges and dilution steps, efficient reagent usage & stability and excellent mean time between failure
Learn more about the Optilite system in the videos below:
"We needed an analyser that could run these special protein assays quickly and efficiently, which led to the decision to move them to the Optilite as a stand alone analyser - The biggest benefit we have seen is to our workflow." 
- Peterborough City Hospital
To deliver the best service to your clinicians, special protein testing requires consistently high-quality results with predictable, short turnaround times. The highly reliable and compact Optilite system features fully automated dilutions plus unrestricted access for samples and reagents, enabling a streamlined workflow for your entire laboratory.
Worldwide, over 1000 satisfied customers are benefiting from the optimised solution of Binding Site assays and systems. Through the support from a global team of technical and scientific protein experts, we bring you consistent throughput and high-quality results combined in a seamless workflow for ultimate peace of mind.
  • Average mean time between failure greater than 19 months*
  • Predictable reporting times enabling optimal management of laboratory staff and resources
  • Full traceability through barcoded reagents, calibrators and controls
  • 3 different methods of antigen excess detection applied when required during assay design
  • IVDR Class A compliant and supports ISO 15189 accreditation
  • Supported by a global team of technical and scientific experts
*Based on 12 months global data between January 2023 and December 2023
Optilite automated benchtop analyzer with silver-gray front panel and reagent compartments for protein testing
  • Consistent and high average throughput of 120 tests/hour 
  • Quick time to reportable result - approximately 40% faster compared to alternative systems1
  • Save reagents with our optimized protocols, including large dilution steps and wide measuring ranges
  • Single-use cuvettes remove risk of cuvette degradation and potential source of carry-over
  • Cost-effective reagent management with up to 3 months open vial stability
  • High-quality test menu including unique assays such as Hevylite® assays, CH50 and IgA1/2
Chart depicting Optilite performance per hour
Click image to enlarge
*75% of samples required at least 1 dilution
Figures based on internal testing                                                                                                                                           
  • Fully automated dilutions for minimal hands-on time
  • Continuous loading and unloading of samples, reagents and disposables
  • Capacity of 360 disposable cuvettes increasing walk-away time
  • Efficient reagent management with on-board cooling
  • Short turnaround time (STAT) function for urgent samples
  • Easy-to-use software and bi-directional laboratory information system (LIS)-interface
Chart depicting efficiency vs cobas
Click image to enlarge
Improvement in Workflow and Efficiency for Measurement of Special Proteins Using the Optilite Standalone Automated Analyzer. Saenger et al, 
Performed at Hennepin County Medical Center, Minneapolis, USA. Presented at the 2021 AACC Annual Scientific Meeting & Clinical Lab Expo, September 26-30, 2021 in Atlanta, Georgia
Read more about the Optilite analyser
The optimal and dedicated analyser for special protein testing
Discover the details
Technical specifications for the Optilite analyser
Why choose the Optilite analyser
Gain in efficiency, productivity and quality
Monoclonal Gammopathies
Freelite® Assay Menu       Product Code       Kit Size      
Freelite Kappa
LK016.OPT
100 tests
Freelite Lambda
LK018.OPT
100 tests
Hevylite® Assay Menu           Product Code      Kit Size     
Hevylite IgG Kappa
NK621.OPT
50 tests
Hevylite IgG Lambda NK622.OPT 50 tests
Hevylite IgA Kappa
NK623.OPT
50 tests
Hevylite IgA Lambda
NK624.OPT
50 tests
Hevylite IgM Kappa
NK625.OPT
50 tests
Hevylite IgM Lambda NK626.OPT 50 tests
Complement
Description            Product Code     Kit Size   
C1 Inactivator Kit NK019.OPT 50 tests
C3c Kit NK023.OPT 100 tests
C4 Kit NK025.OPT 100 tests
CH50 Reagent NK095.OPT 100 tests
CH50 Calibrator NC095.OPT 1 pack
CH50 Controls NQ095.OPT 1 pack
Central nervous system disorders
Description                         Product Code       Kit Size  
Freelite Mx kappa Latex Kit LK016.M.OPT 100 tests
Freelite Mx™ Lambda Latex Kit     LK018.M.OPT          100 tests
Low Level Albumin Kit NK032.L.OPT 100 tests
Low Level IgG Kit NK004.L.OPT 60 tests
IgA CSF Kit LK010.L.OPT 60 tests
IgM CSF Kit LK012.L.OPT 60 tests
Immunoglobulins
Immunoglobulin assays     Product Code   Kit Size  
IgG Kit NK004.OPT 100 tests
IgA Kit NK010.OPT 100 tests
IgM Kit NK012.OPT 100 tests
IgD Kit LK013.OPT 100 tests
IgE Kit* LK014.OPT 100 tests
Renal Function
Description                  Product Code   Kit Size  
Cystatin C LK048.OPT 100 tests
Albumin NK032.OPT 100 tests
Low Level Albumin NK032.L.OPT 100 tests
Beta-2-Microglobulin LK043.OPT 100 tests
Low Level IgG NK004.L.OPT 60 tests
Transferrin Urine NK070.U.OPT 100 tests
Beta-2-Microglobulin Urine LK043.L.OPT 100 tests
Alpha-1-Microglobulin Urine NK036.U.OPT 50 tests
Alpha-2-Macroglobulin Urine   NK039.U.OPT 100 tests
Subclasses
Description Product Code Kit Size
IgG1 Kit NK006.OPT 100 tests
IgG2 Kit NK007.OPT 100 tests
IgG3 Kit   LK008.OPT 100 tests
IgG4 Kit  
LK009.OPT
100 tests
IgA1 Kit NK087.OPT 50 tests
IgA2 Kit LK088.OPT 50 tests
Specific proteins
Description Product Code Kit Size
Alpha-1-Acid Glycoprotein Kit NK063.OPT 100 tests
Alpha-1-Antitrypsin Kit NK034.OPT 100 tests
Alpha-2-Macroglobulin Kit NK039.OPT 100 tests
Anti-Streptolysin O Kit LK189.OPT 100 tests
Anti-Tetanus Toxoid Immunoglobulin Kit LK110.OPT 200 tests
Apolipoprotein A-1 Kit* NK085.OPT 100 tests
Apolipoprotein B Kit* NK086.OPT 100 tests
Caeruloplasmin Kit NK045.OPT 50 tests
C-Reactive Protein Kit* NK044.OPT 100 tests    
High Sensitivity C-Reactive Protein Kit           LK044.L.OPT 100 tests
Haptoglobin Kit NK058.OPT 100 tests
Lipoprotein Kit* LK098.OPT 100 tests
Prealbumin Kit NK066.OPT 100 tests
Rheumatoid Factor Kit LK151.OPT 100 tests
Total Protein Kit* NK061.OPT 100 tests
Transferrin Kit NK070.OPT 100 tests
Vaccine response
Description Product Code    Kit Size  
Tetanus Toxoid IgG response MK010 96 tests
Diptheria Toxoid IgG response MK014 96 tests
Haemophilus influenzae type B IgG MK016 96 tests
Pneumococcal Capsular Polysaccharide (PCP) IgG response   MK012 96 tests
Salmonella typhi Vi IgG  response MK091 96 tests
*Calibrators and Controls are supplied separately
Product availability is subject to specific regulatory requirements.
Optimized immunoassays for the Optilite® Analyser
Immunoassays
Optimized immunoassays for the Optilite® Analyser
Binding Site Immunoassays are optimised for use on the Optilite System for consistent and high-quality results. Learn about our extensive range of Immunoassays available for special protein analysis.
Peer to peer QC comparison with LabLink xL™ software
LabLink xL software
Peer to peer QC comparison with LabLink xL™ software
LabLink xL software is the dedicated peer QC solution for Binding Site controls run on the Optilite System. LabLink xL software allows the laboratory to monitor their mean values for precision and accuracy within a peer-group using the same analyzer, and the same lot of the same assay kit.
References
  1. Ghillani P, et al. Analytical performances of Optilite® turbidimeter (The Binding Site): a new dedicated analyser for specific proteins determination. Ann Biol Clin (Paris) 2017; 75:29-37
Product availability is subject to specific regulatory requirements.
Contact your local representative for availability in your country. Optilite, Freelite and Hevylite are registered trademarks of the Binding Site Group Ltd (Birmingham, UK) in certain countries. Freelite Mx is a trademark of the Binding Site Group Ltd (Birmingham, UK) in certain countries. Other Brand or product names may be trademarks of their respective holders.
Contact Us?
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you