Simplified workflow for special protein testing
"We needed an analyser that could run these special protein assays quickly and efficiently, which led to the decision to move them to the Optilite as a stand alone analyser - The biggest benefit we have seen is to our workflow."
- Peterborough City Hospital
To deliver the best service to your clinicians, special protein testing requires consistently high-quality results with predictable, short turnaround times. The highly reliable and compact Optilite system features fully automated dilutions plus unrestricted access for samples and reagents, enabling a streamlined workflow for your entire laboratory.
Worldwide, over 1000 satisfied customers are benefiting from the optimised solution of Binding Site assays and systems. Through the support from a global team of technical and scientific protein experts, we bring you consistent throughput and high-quality results combined in a seamless workflow for ultimate peace of mind.
*75% of samples required at least 1 dilutionFigures based on internal testing |
Improvement in Workflow and Efficiency for Measurement of Special Proteins Using the Optilite Standalone Automated Analyzer. Saenger et al,Performed at Hennepin County Medical Center, Minneapolis, USA. Presented at the 2021 AACC Annual Scientific Meeting & Clinical Lab Expo, September 26-30, 2021 in Atlanta, Georgia |
| Freelite™ Assay Menu | Product Code | Kit Size |
|---|---|---|
| Freelite Kappa |
LK016.OPT.A |
100 tests |
| Freelite Lambda |
LK018.OPT.A |
100 tests |
| Hevylite™ Assay Menu | Product Code | Kit Size |
|---|---|---|
| Hevylite IgG Kappa |
NK621.OPT.A |
50 tests |
| Hevylite IgG Lambda |
NK622.OPT.A |
50 tests |
| Hevylite IgA Kappa |
NK623.OPT.A |
50 tests |
| Hevylite IgA Lambda | NK624.OPT.A | 50 tests |
| Hevylite IgM Kappa |
NK625.OPT.A |
50 tests |
| Hevylite IgM Lambda | NK626.OPT.A | 50 tests |
| Description | Product Code | Kit Size |
|---|---|---|
| C1 Inactivator Kit | NK019.OPT.A | 50 tests |
| C3c Kit | NK023.OPT.A | 100 tests |
| C4 Kit | NK025.OPT.A | 100 tests |
| CH50 Reagent | NK095.OPT.A | 100 tests |
| CH50 Calibrator | NC095.OPT.A | 1 pack |
| CH50 Controls | NQ095.OPT.A | 1 pack |
| Description | Product Code | Kit Size |
|---|---|---|
| Low Level Albumin Kit | NK032.L.OPT.A | 100 tests |
| Low Level IgG Kit | NK004.L.OPT.A | 60 tests |
| IgA CSF Kit | LK010.L.OPT.A | 60 tests |
| IgM CSF Kit | LK012.L.OPT.A | 60 tests |
| Immunoglobulin assays | Product Code | Kit Size |
|---|---|---|
| IgG Kit | NK004.OPT.A | 100 tests |
| IgA Kit | NK010.OPT.A | 100 tests |
| IgM Kit | NK012.OPT.A | 100 tests |
| IgD Kit | LK013.OPT.A | 100 tests |
| Description | Product Code | Kit Size |
|---|---|---|
| Albumin | NK032.OPT.A | 100 tests |
| Beta-2-Microglobulin | LK0043.OPT.A | 100 tests |
| Beta-2-Microglobulin Urine | LK043.L.OPT.A | 100 tests |
| Cystatin C | LK048.OPT.A | 100 tests |
| Low Level Albumin | NK032.L.OPT.A | 100 tests |
| Low Level IgG | NK004.L.OPT.A | 60 tests |
| Description | Product Code | Kit Size |
|---|---|---|
| IgG1 Kit | NK006.OPT.A | 100 tests |
| IgG2 Kit | NK007.OPT.A | 100 tests |
| IgG3 Kit | LK008.OPT.A | 100 tests |
| IgG4 Kit |
LK009.OPT.A |
100 tests |
| IgA1 Kit | NK087.OPT.A | 50 tests |
| IgA2 Kit | LK088.OPT.A | 50 tests |
| Description | Product Code | Kit Size |
|---|---|---|
| Alpha-1-Acid Glycoprotein Kit | NK063.OPT.A | 100 tests |
| Alpha-1-Antitrypsin Kit | NK034.OPT.A | 100 tests |
| Alpha-2-Macroglobulin Kit | NK039.OPT.A | 100 tests |
| Anti-Streptolysin O Kit | LK189.OPT.A | 100 tests |
| Caeruloplasmin Kit | NK045.OPT.A | 50 tests |
| C-Reactive Protein Kit* | NK044.OPT.A | 100 tests |
| Haptoglobin Kit | NK058.OPT.A | 100 tests |
| Prealbumin Kit | NK066.OPT.A | 100 tests |
| Rheumatoid Factor Kit | LK151.OPT.A | 100 tests |
| Transferrin Kit | NK070.OPT.A | 100 tests |
| Description | Product Code | Kit Size |
|---|---|---|
| Tetanus Toxoid IgG response | MK010 | 96 tests |
| Diptheria Toxoid IgG response | MK014 | 96 tests |
| Haemophilus influenzae type B IgG | MK016 | 96 tests |
| Pneumococcal Capsular Polysaccharide (PCP) IgG response | MK012 | 96 tests |
| Salmonella typhi Vi IgG response | MK091 | 96 tests |