Specific protein assays
Your partner in targeted protein analysis
Introduction
What are Specific Protein assays?
Specific protein assays determine the amount or concentration of a specific protein or an array of different proteins in a sample.
Different assays in this group may aid in the characterisation of a number of disorders.  Individual assays in this group may aid in the characterization of:
  • Auto-immune disorders
  • Infections
  • Hemolytic diseases (anemias)
  • Metabolic disorders and other diseases
Optilite™ analyzer provides automated specific protein assays
with enhanced efficiency and optimized workflow.
Range of assays
The following assays are included in the specific protein assay range
Inflammation assays
  • A1 Acid Glycoprotein
  • A1 Antitrypsin
  • ASO
  • CRP
  • Rheumatoid factor
Other assays
  • Caeruloplasmin
  • Haptoglobin
  • Prealbumin
  • Transferrin
Abbreviation: ASO - Antistreptolysin O; CRP – C-reactive protein 
  • Anti-Streptolysin O (ASO)- Can be used to establish the degree of infection past or present, by β-haemolytic streptococci
  • Rheumatoid Factor (RF)- A marker that may aid in the characterisation of rheumatoid arthritis 
  • ɑ1 Acid Glycoprotein & C Reactive Protein are acute phase reactants raised in acute & chronic inflammatory conditions 
  • ɑ1 Antitrypsin deficiency is associated with conditions like pulmonary emphysema
  • Anti-Streptolysin O (ASO)- Can be used to establish the degree of infection past or present, by β-haemolytic streptococci
  • Rheumatoid Factor (RF)- A marker that may aid in the characterisation of rheumatoid arthritis 
  • ɑ1 Acid Glycoprotein & C Reactive Protein are acute phase reactants raised in acute & chronic inflammatory conditions 
  • ɑ1 Antitrypsin deficiency may lead to conditions like pulmonary emphysema
  • High sensitive C-reactive protein (hs CRP) can aid in identification of inflammation
  • Haptoglobin- A diagnostic marker for hemolytic diseases such as anemia, liver disease and inflammatory conditions
  • Caeruloplasmin- A diagnostic marker for copper metabolism disorders, such as Menkes or Wilson’s disease
  • Prealbumin- Aids in assessment of nutritional status 
Photo of an Optilite analyser in focus with the words "optimised protein system" under the logo.
Product
Optilite Analyser
Transform your diagnostic testing with the Optilite™ Analyser
Photo of an operator loading samples in to the Optilite analyser
Product
Freelite™ Assays
Detect and Monitor Multiple Myeloma with Freelite™ assays
Contact Us?
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you
References
  1. Nordestgaard, B.G., Chapman, M.J., Ginsberg, H.N., The European Atherosclerosis Society Consensus Panel. (2012). Lipoprotein(a): EAS recommendations for screencing, desirable levels and management