Overview
Immunoglobulins are glycoproteins consisting of heavy and light chains produced by plasma cells that mediate humoral immunity by producing IgM, IgD, IgG, IgA and IgE1,2.
Immunoglobulins comprise approximately 20% of the protein in plasma.1
Quantitative analysis of serum immunoglobulins is considered a first-line diagnostic measure for primary immunodeficiency.
There are five classes of antibodies, each having a slightly different structure:
These structural differences give them different functions and allow them to be found in different locations in the body.
Immunodeficiency testing
Binding Site provides automated solutions for the measurement of IgA, IgD, IgG and IgM on the Optilite™ system.
Over 30 years of experience in immunodeficiency testing