EXENT™ GAM Assay
The Immunoglobulin Isotypes (GAM) for the EXENT™ Analyser (EXENT™ GAM Assay) aids in confirmatory diagnosis of monoclonal gammopathies
Overview
What is the EXENT GAM Assay?
The EXENT GAM Assay is a sensitive and specific immunoassay that identifies and quantifies M-proteins based on their unique molecular mass. Sample preparation uses paramagnetic beads specific to human IgG, IgA, IgM, total Kappa and total Lambda to isolate immunoglobulins from patient serum. The isolated immunoglobulins are analyzed by MALDI-ToF mass spectrometry to specifically measure the monoclonal protein component. The assay is used in conjunction with the Binding Site Optilite™ IgG, IgA and IgM assays to provide semi quantification / reporting in g/L of identified monoclonal proteins.​
A new paradigm for the detection & isotyping of M-proteins
Monoclonal protein detection using mass spectrometry
Plasma cell clone
Each plasma cell produces a unique immunoglobulin
Plasma cell clone
Immunoglobulin molecules consist of two identical heavy chains and two identical light chains (Kappa or Lambda)
Plasma cell clone
When immunoglobulins have their inter-chain disulphide bonds reduced, they are dissociated into their constituent heavy and light chains. The reduced light chains from the intact immunoglobulins are ideally suited to analysis by MALDI-ToF mass spectrometry due to their low molecular weight
Plasma cell clone
The resulting mass spectra reflects the plot of mass vs intensity of the isolated and reduced immunoglobulins
(A) When immunoglobulins are isolated from the serum of a healthy individual, by specific immunopurification, the resulting MALDI-ToF mass spectrometry resolves three normally distributed peaks that are representative of the polyclonal Lambda, Kappa and heavy Kappa free light chains derived from the intact immunoglobulins.1                         
(B) In a patient with a monoclonal gammopathy, the presence of a sharp peak within the polyclonal background is indicative of the presence of a monoclonal immunoglobulin. The molecular weight of the identified peak can be used to specifically track the monoclonal protein and the peak area ratio of the monoclonal to polyclonal can be combined with the Optilite IgG, IgA or IgM assay to report a result in g/L.                     
When a monoclonal protein is identified, a simple calculation is applied to provide a test result in g/L based on the total immunoglobulin value derived from the Optilite IgG, IgA or IgM assay. ​
The EXENT GAM Assay can be used to:
  • Aid in evaluation of monoclonal gammopathy of undetermined significance (MGUS)
  • Aid in the diagnosis of smoldering multiple myeloma (SMM), multiple myeloma (MM), waldenström's macroglobulinemia (WM) and AL amyloidosis
The assay empowers clinicians to effectively diagnose patients with monoclonal gammopathies with a simple serum test. This offers the opportunity to:
  • Confirm M-protein and M-protein isotype at diagnosis with enhanced sensitivity when serum protein electrophoresis suggests an M‑protein or when serum free light chains are abnormal2
  • Identify monoclonal proteins based on their unique molecular mass, providing reduced ambiguity in result interpretation
  • Have diagnostic confidence and disease clarity through clear and objective results, with automated peak picking
  • Identify clinically relevant information such as post translational modifications and secondary clones
Related content
EXENT System
The EXENT System is an automated solution that enables M-protein identification and measurement in serum with enhanced sensitivity beyond conventional methods
This product is not commercially available in China. Future commercial availability cannot be guaranteed.
EXENT and Optilite are registered trademarks of The Binding Site Group Limited (Birmingham, UK) in certain countries.
The EXENT™ System combines EXENT™ Analyser with Immunoglobulin Isotypes (GAM) for the EXENT™ Analyser.
EXENT GAM Assay refers to Immunoglobulin Isotypes (GAM) for the EXENT™ Analyser.
Contact Us?
Questions about our products or services? Our team is ready to hear them.
Complete this form and we will get back to you
References
  1. Barnidge DR, et al. Subset of kappa and lambda germline sequences result in light chains with a higher molecular mass phenotype. J Proteome Res 2015; 14:5283-90
  2. Immunoglobulin Isotypes (GAM) for the EXENT Analyser Instructions for Use