Events
ECTRIMS 2025: Stepping into a new era of precision in MS Diagnosis
For the fourth year in a row, I had the privilege of representing the Binding Site at the European Committee for Treatment and Research in Multiple Sclerosis (ECTRIMS). This year, ECTRIMS, the biggest neurology conference in the world, was held in Barcelona. With more than 9,600 participants from 120 countries and 2,513 abstracts, this was the largest ECTRIMS to date.
New guidelines took center stage
The program revolved around the theme "A new era of precision," aligning perfectly with the newly released 2024 McDonald criteria1. The new revision of the guidelines for the diagnosis of Multiple Sclerosis (MS) was published just a few days before the conference started.
When first presented at ECTRIMS 2024, Professor Tintoré, MD, PhD, described this revision as "the most diverse, gender-balanced, and country-balanced diagnostic criteria developed to date." 2  I found it striking how quickly these new guidelines have moved from theory to practice. Session after session, real-world data3,4 highlighted how the guidelines enable earlier diagnosis. By providing new and accessible tools to aid in the diagnosis of MS, the revised McDonald criteria offer a pragmatic approach tailored to what clinicians can actually request locally.
The Kappa Free Light Chain Index (κFLC index) is one of the new updates to the McDonald criteria. Other changes include the use of the optic nerve as the fifth anatomical location to be examined for lesions, and the detection of Central Vein Sign (CVS) and Paramagnetic Rim Lesions (PRLs). 
Kappa FLC Index: accelerating MS diagnosis
Among the updates to the McDonald criteria, a new biomarker has been included: the Kappa FLC Index. This can now be used as an alternative to the traditional OCB method1. According to the 2024 McDonald criteria, a Kappa FLC Index above 6.1 is defined as 'CSF positive'1.
Kappa FLC Index allows the characterization of intrathecal immunoglobulin synthesis, a hallmark of MS. In addition, it offers several advantages over OCB testing, including being quantitative, fast, an easy-to perform and automated technique, with rater-independent interpretation. OCB testing, on the other hand, is time-consuming and technically demanding, labor intensive and, due to its qualitative results, may suffer of rater-dependent interpretation5.
Compared to last year’s ECTRIMS, neurologists weren’t just curious about the Kappa FLC Index. They are now ready to request it and implement it in their routine practice.
ECTRIMS 2025, in the wake of the 2024 McDonald criteria, effectively moved the Kappa FLC Index from "scientifically validated" to "ready for routine use."
Expert insights on κFLC Index
I found the "Meet the Expert" 6 session chaired by Professor Arrambide, MD, PhD and Professor Moccia, MD, PhD particularly enlightening, as it made the practical case for the use of Kappa FLC Index crystal clear.
When asked which technique the speakers would recommend if a laboratory must choose between OCB or Kappa FLC Index testing, Prof. Moccia replied without hesitation. "The Kappa FLCs.", he stated, while Prof. Arrambide nodded enthusiastically. He continued, "It’s cheaper, it’s quicker, and it provides a quantitative number."
Prof. Arrambide went on to talk about OCB testing: "In-house tests are much better for detecting oligoclonal bands than commercial ones. Commercial [OCB tests] are better than nothing, but in that case, it might be better to start with the Kappa FLC." 
She then added, "There are at least a couple of studies looking at costs, and yes, as Marcello [Prof. Moccia] said, they [κFLCs] are cheaper."
A key point from this session was that κFLC index accelerates result availability. Results can be reported in 20–30 minutes, and running the test does not require special training.
The scientific community’s enthusiasm for the CSF κFLC analysis and Index was also evident during the poster sessions. With a record number of researchers presenting their work, more than 20 studies investigated the use of the Kappa FLC Index in MS and other neurological diseases.
Access to new paraclinical tests
I appreciated the honesty of the multi-country surveys regarding global access to the new paraclinical tools included in the 2024 McDonald criteria, including κFLC Index. Access is currently uneven, and there’s work to be done7,8. The barriers are familiar: reimbursement, training gaps, and translation of international guidelines into national consensus7.
On the other hand, several presenters argued that the obstacles to availability of κFLC testing may be perceived rather than real. In various sessions, experts highlighted that hematology laboratories already run FLC assays for Multiple Myeloma investigations. Within this situation, adding CSF κFLC testing becomes straightforward. The real barrier may therefore not be a lack of availability of this technology, but awareness. Enhanced collaboration between departments could be key to accessing Kappa FLC testing6.
A practical takeaway I loved: if you need κFLC Index, ask your hematology lab6.
Conclusions
ECTRIMS 2025 was important to me because it marked the point where precision in MS diagnosis became truly operational. I saw the community shift from discussing the evidence, to implementing it and embracing tools that are faster and more accessible.
Watching the κFLC Index progressing from validation to routine use, supported by guideline inclusion and real-world feasibility, was deeply motivating. 
References
  1. Montalban X, et al. Diagnosis of multiple sclerosis: 2024 revisions of the McDonald criteria. Lancet Neurol 2025; 24:850-865
  2. ECTRIMS. Scientific Session 1: New diagnostic criteria. 2024  [09/2024]; Available from: https://apps.congrex.com/ectrims2024/en-GB/pag/session/65077.
  3. Bollo L, et al. Impact of the 2024 McDonald criteria revisions on multiple sclerosis diagnosis: a comparison with the 2017 criteria. Presented at ECTRIMS 2025; O001a
  4. Brownlee W, et al. Real-world validation of the 2024 McDonald criteria in patients under evaluation for suspected multiple sclerosis. Presented at ECTRIMS 2025; O002a
  5. Hegen H, et al. Cerebrospinal fluid kappa free light chains for the diagnosis of multiple sclerosis: A consensus statement. Mult Scler 2022; 29:182-195
  6. ECTRIMS. Meet the Expert 4: Applications of 2024 diagnostic criteria for MS: implications for the use of MRI, OCT, and body fluid biomarkers. 2025  [09/2025]; Available from: https://pag.virtual-meeting.org/congrex/ectrims2025/en-GB/pag/session/67958.
  7. Solomon A, et al. Global Access to Paraclinical Diagnostic Testing for Multiple Sclerosis: Results of the 2024 Multiple Sclerosis International Federation Atlas of MS Topical Survey. Presented at ECTRIMS 2025; O003a
  8. ECTRIMS. Scientific Session 16: Global views - diagnostic criteria across the world. 2025  [09/2025]; Available from: https://pag.virtual-meeting.org/congrex/ectrims2025/en-GB/pag/session/67949.