FTIR Basic Organic Functional Group Reference Chart (PDF)
Functional groups are structural units within organic compounds defined by specific atom and bond arrangements. Infrared is a powerful identification tool for functional groups because of the similar absorption frequencies for those groups in different molecules. The actual frequency is affected by the environment, so the reference chart shows wide bands rather than specific frequencies. The identification of functional groups is a cornerstone of IR spectroscopy and organic chemistry.
To help make your job easier, we outlined the basic organic functional groups, their positions of infrared bans, their relative peak intensities, and their functional group representation. Download this easy-to-read chart.
Looking for more basic FTIR information, be sure to visit our FTIR Spectroscopy Learning Center and visit the Introduction and Basics page.
very useful
sir ,
i want to analys my IR spectra peaks , that i will give here, cn i gets its results of analysis.
Peak At
3946.5973
3865.2780
3847.9582
3826.7419
3778.6400
3749.3190
3689.4888
2864.1387
2387.3082
2339.7518
2254.5750
2231.1183
2202.8104
2177.5028
2148.9612
2131.2128
2090.5921
1963.4113
1930.3108
1874.7277
1842.2506
1784.5439
1545.4176
1484.9249
1408.1645
1328.8908
1304.5184
1270.8724
1178.9547
902.2080
846.9365
802.3415
737.0366
597.8158
576.7164
462.8811
446.7886
418.7146
pls analyse such peaks
ok
hi sir
i sent an emial for my result that need to anaylsis
Ravindra,
I cannot answer your question as given here. I would need either the raw data or a picture to even begin, as the large number of peaks you list is very confusing. A look at the spectrum would be the only way to help you.
Please I do have some FTIR scanned results/peaks i would like to analyse, please do reach out to me via skygold12@gmail.com so i can forward them to you please.
thanks
Kindly assist me with the full analysis of these FTIR results.
you can reach me skygold12@gmail.com so i can send over the pictures of the Scan/pictures.
thanks
Won’t download. Says I don’t have permission
I have FTIR analysis peaks with me Please help me to identify the functional groups .
this really help