Antidoping detection strategies to combat emerging threats
Robust doping control measures are essential to protect the belief that victory is earned through hard work, talent, and dedication, rather than unethical shortcuts. Utilizing highly sensitive and high-resolution analytical technologies is paramount in the fight against doping in sports. These advanced analytical tools help enable detection of even minuscule traces of prohibited substances, leaving minimal opportunity for undetected doping.
World Anti-Doping Agency: Celebrating 25 years
Prohibited substance screening
Initial testing procedures (ITPs) efficiently screen urinary or blood samples for numerous prohibited substances, at times requiring high-resolution mass spectrometry with resolution over 60,000 FWHM and mass errors under 2 ppm. These assays for direct urine analysis target substances such as steroids, stimulants, corticoids, metabolic modulators, diuretics, narcotics, cannabinoids, and low molecular mass peptides.
The table below provides a general overview of doping control analysis including the classes of prohibited compounds and the mainly applied methodology.
LC-MS, LC-MS/MS, LC-HRAM MS GC-MS or GC-MS/MS Alternative analysis
Glucocorticoids
Stimulants
Anabolic agents
β2-agonists
β-blocker
Narcotics
Metabolic modulators
Diuretics
Peptides (small & large)
Proteases
siRNA
Exogenous steroids
Endogenous steroids
Stimulants
Narcotics
β2-agonists
Cannabinoids
Anabolic agents
Xenon
Erythropoiesis stimulating agents
Growth hormone (GH)
Chorionic gonadotrophin (CG) and luteinizing hormone (LH)
Hematological parameters
Blood transfusion
Gene doping
Initial testing procedures (ITP)
Confirmation procedures (CP)
Initial testing procedures (ITP)
Confirmation procedures (CP)
Initial testing procedures (ITP)
Confirmation procedures (CP)
High-Resolution MS: Orbitraps, time-of-flight MS
LC-MS/MS: triple-quadrupoles, linear ion traps
Ionization: ESI, APCI
GC-MS: single quadrupoles
GC-MS/MS: triple quadrupoles
GC-C-IRMS: single magnetic sector analyzer
Ionization: EI
ECLIA
IEF, SDS-SAR-PAGE
Flowcytometry
ILMA
PCR
*Abbreviations: GC-MS: gas chromatography mass spectrometry, LC-MS: liquid chromatography mass spectrometry, HRMS: high-resolution mass spectrometry, IRMS: isotope-ratio mass spectrometry, C: combustion, EI: electron ionization, ESI: electro spray ionization, APCI: atmospheric pressure chemical ionization, SDS-PAGE: sodium dodecyl sulfate-polyacrylamide gel electrophoresis, ECLIA: electrochemiluminescence immunoassay, IEF: isoelectric focusing, SAR: sarcosyl, ILMA: immunoluminometric assay, PCR: polymerase chain reaction.
“Every variable of an analytical assay is optimized, from the selection of the sample matrix to the ideal purification and concentration strategy, to the best possible analyte separation approach, to the most specific and sensitive analyzer. The chances of mastering this endeavor are substantially improved by innovations in separation science and mass spectrometric instrumentation. Here, close collaboration in research and development with industry partners such as Thermo Fisher Scientific has proven to be invaluable in achieving the goals of sports drug testing programs.”
Mario Thevis
Director, Center for Preventative Doping Research
German Sport University Cologne
Typical prohibited substance screening workflows
Solid phase extraction (SPE) is an effective and versatile extraction technique for small peptides and other prohibited substances associated with the field of doping. Starting with urine-based samples, laboratories will need to utilize a multi-step preparation to maximize system sensitivity.
Thermo Scientific HyperSep Hypercarb SPE Cartridges
  • SPE cartridge retains highly polar compounds
  • Unique graphite carbon-based material
  • pH stable 0 – 14
Thermo Scientific SOLA SPE Cartridges
  • Greater throughput with reduced solvent requirements
  • Cleaner extracts for higher confidence results
Videos
Targeted screening focuses on identifying specific known substances, such as anabolic steroids, stimulants, or erythropoiesis-stimulating agents, using pre-defined analytical methods like liquid chromatography-mass spectrometry (LC-MS) or gas chromatography-mass spectrometry (GC-MS). Over the past decade, the majority of targeted screening of the initial testing procedures (ITP) utilized in testing labs have employed high-resolution analytical techniques to capture a wide range of compounds, including new designer drugs or unusual biomarkers of doping. An independent, confirmatory analysis of the suspicious sample follows and is focused solely on the prohibited substances and its metabolite.
Thermo Scientific
TSQ 9610 Triple Quadrupole GC-MS
Thermo Scientific TSQ Altis Plus Triple Quadrupole Mass Spectrometer
Thermo Scientific
Orbitrap Exploris 120 Mass Spectrometer*
  • Sensitive quantitation meets updated MRPLs
  • He carrier gas saving technology
  • NeverVent technology for planned maintenance
  • Low-level compound detection and quantitation
  • 800 SRM/sec for fast, routine quantitation
  • 5ms polarity switching
  • Employed for confirmatory analysis
  • Orbitrap analyzer increases resolving power
  • LC-HRAM MS system-to-system reproducibility
  • Max compound coverage with fast scan rates
  • Employed for prohibited substance screening
*The Thermo Scientific Orbitrap Exploris 240 may also be used for prohibited substance screening.
Thermo Scientific TraceFinder data acquisition and processing software is widely used for the analysis of doping compounds, offering streamlined data processing and high-throughput screening capabilities. It helps enable targeted detection by integrating LC-MS data with automated peak identification, quantification, and reporting. As an alternative, Thermo Scientific Chromeleon Chromatography Data System Software may also be employed.
Thermo Scientific TraceFinder Software
  • Complete screening and quantitation software
  • Driven by powerful, intuitive acquisition and data processing
  • Smart sample flagging, flexible data review, and custom report generation removes bottlenecks associated with data review
Thermo Scientific Chromeleon Chromatography Data System (CDS) Software
  • Supports targeted quantitation and screening MS workflows
  • Compliance and data integrity tools as standard
  • Improved sharing and security of data without need for a separate server
Doping control with retrospective analysis
High-resolution accurate mass (HRAM) mass spectrometry (MS) has become the technology of choice for precise mass measurements and the ability to detect new doping markers. Widely used in doping analysis, laboratories trust Thermo Scientific proprietary Orbitrap mass spectrometers, which are capable of running thousands of sample injections for the comprehensive screening of both known and unknown performance-enhancing substances.
High-resolution Orbitrap MS enables retrospective analysis of stored samples to confirm new illicit substances. The International Testing Agency (ITA) stores athlete samples for 10 years, deterring misuse of performance-enhancing substances. Combined with retrospective analysis, it helps detect new metabolites, extend detection windows, and monitor an athlete's biological passport (ABP) for longitudinal profiling.
“Thinking back to 2012, I recall that our aim was for our methods to be “super-fast and super sensitive”. Using the Q Exactive Orbitrap mass spectrometer in positive, negative and all ion fragmentation modes in 1.3 seconds for non-targeted data capture met our aim and was certainly a first at any Olympics meeting.”
David Cowan
Former Director of the London Drug Control Centre
Dept. of Analytical, Environmental & Forensic Sciences
King’s College London
Typical doping control with retrospective analysis workflows
Solid phase extraction (SPE) is an effective and versatile extraction technique for small peptides and other prohibited substances associated with the field of doping. Starting with urine-based samples, laboratories will need to utilize a multi-step preparation to maximize system sensitivity.
Thermo Scientific HyperSep Hypercarb SPE Cartridges
  • SPE cartridge retains highly polar compounds
  • Unique graphite carbon-based material
  • pH stable 0 – 14
Thermo Scientific SOLA SPE Cartridges
  • Greater throughput with reduced solvent requirements
  • Cleaner extracts for higher confidence results
Videos
Thermo Scientific Orbitrap Exploris GC 240 Mass Spectrometer
  • Performance and accuracy—240,000 mass resolving power, MS/MS capability, and high sensitivity
  • Discover with ease—Intuitive instrument control and method templates
  • Versatility—Easily switch between techniques, ion sources, probes, and columns
Thermo Scientific Vanquish Flex UHPLC coupled to Orbitrap Exploris 240 Mass Spectrometer
  • Single channel UHPLC
  • Accelerate qualitative and quantitative confidence with consistently accurate data for targeted and untargeted analysis
  • Retrospective analysis to find novel psychoactive substances as the emerge
  • Resolution up to 120,000 to 240,000 (FWHM) at m/z 200
Thermo Scientific TraceFinder Software
  • Complete screening and quantitation software
  • Driven by powerful, intuitive acquisition and data processing
  • Smart sample flagging, flexible data review, and custom report generation removes bottlenecks associated with data review
Thermo Scientific Compound Discoverer Software
  • For untargeted screening, identify unknowns faster with connection to online mzCloud 2.0 spectral libraries or local Thermo Scientific mzVault spectral libraries
  • Fully integrated suite of advanced tools for both known-parent and unknown data processing and interpretation
  • Streamlines compound identification, comparative analyses
  • Provides extensive filtering and data visualization capabilities
Steroid quantitation and confirmation
Steroid quantitation and confirmation are critical in doping control to detect the misuse of anabolic steroids, corticosteroids, and other performance-enhancing substances. Analytical instrumentation such as mass spectrometers play a vital role in helping to ensure the accuracy, sensitivity, and reliability of these detections.
GC-triple quadrupole (QQQ) MS and GC-high resolution accurate mass MS continue to play a crucial role in antidoping laboratories, especially for analyzing (naturally occurring) androgens in steroid profile studies and detecting anabolic androgenic steroids (AAS) and their metabolites. Additionally, GC coupled with isotope ratio or combustion isotope ratio mass spectrometry (GC/C/IRMS) enhances sensitivity, distinguishing natural from synthetic steroids.
Athlete Biological Passport testing
Antidoping laboratories use two main strategies. The first involves analyzing a single sample, typically at competitive events, to detect prohibited substances. The second is longitudinal testing, where results are part of a broader performance profiling framework. This approach forms the basis of the Athlete Biological Passport (ABP), which monitors the effects of doping on the body over time, which allows anti-doping organizations to establish individual baseline profiles for each athlete. By tracking fluctuations outside an athlete's normal range, the ABP helps identify potential doping practices, including steroid abuse.
Endogenous anabolic androgenic steroids testing workflow
Screening for anabolic steroid misuse and its metabolites in athletes is essential to maintaining integrity in sports. Accurate quantification of endogenous versus exogenous steroids requires consistent sample preparation. Thermo Fisher Scientific provides advanced sample preparation solutions for steroid analysis using highly sensitive techniques like mass spectrometry.
Thermo Scientific SOLAμ SPE Plates
  • Reproducible, robust sample extraction using very low samples and solvent volumes
  • Award-winning innovative frit-less SPE technology
  • Low analytical sample failures
Anabolic-androgenic steroids (AAS) remain prevalent in doping, despite known health risks, including hormonal, cardiovascular, and psychological effects. In 2022, AAS accounted for over 86% of anabolic agent-related Adverse Analytical Findings (AAFs), highlighting the need for improved antidoping testing. Mass spectrometers enable the identification of adverse analytical findings (AAF), which occur when prohibited substances or their markers are found in an athlete's sample, indicating a potential violation of antidoping regulations.
Thermo Scientific Vanquish High-throughput LC Systems
Utilize Thermo Scientific Vanquish High-throughput LC Systems to improve productivity and save bench space. Vanquish Tandem LC Systems increase sample throughput and MS utilization for LC-MS applications by eliminating column washing and equilibration steps from method cycle time. Vanquish Duo UHPLC Systems combine two completely independent LC flow paths to double sample throughput or run independent analyses in parallel.
Thermo Scientific TSQ 9610 Triple Quadrupole GC-MS
Thermo Scientific TSQ Altis Plus Triple Quadrupole Mass Spectrometer
Thermo Scientific Orbitrap Exploris 120 Mass Spectrometer
Thermo Scientific Stellar Mass Spectrometer
  • Sensitive quantitation meets updated MRPLs
  • He carrier gas saving technology
  • NeverVent technology for planned maintenance
  • Efficient Q2 ion transmission for low mass range
  • Faster Pos/Neg switching times
  • Improved spectral quality and long-term temperature stability
  • Orbitrap analyzer increases resolving power
  • LC-HRAM MS system-to-system reproducibility
  • Max compound coverage with fast scan rates
  • Expanded scale, higher sensitivity, greater specificity, increased throughput
  • HCD and CID fragmentation ability and 5 ms polarity switching
  • Scan rates for MS2 and MS3 of 124 and 40 Hz, respectively
TraceFinder data acquisition software is widely used for the analysis of doping compounds, including steroids, offering streamlined data processing and high-throughput screening capabilities. It facilitates both targeted and untargeted detection by integrating multi-channel LC, single quadrupole, triple quadrupole and high-resolution LC-MS systems, as well as IC and GC systems.
Thermo Scientific TraceFinder Software
  • Complete screening and quantitation software
  • Driven by powerful, intuitive acquisition and data processing
  • Smart sample flagging, flexible data review, and custom report generation removes bottlenecks associated with data review
GC coupled with isotope ratio MS or combustion isotope ratio MS (GC/C/IRMS) allows distinguishing natural and synthetic steroids by measuring carbon isotope ratio. A classic example is testosterone—both an endogenous compound in the body and an exogenous anabolic androgenic steroid. The analytical measurement of the 13 C/12 C ratio can distinguish between the endogenous steroid production versus synthetic administration of the steroid.
Thermo Scientific GC IsoLink II IRMS System
  • Unique and fully automated combustion and high-temperature conversion unit for compound-specific analysis of 13C, 15N, 18O, and 2H
  • Sharp peak shape and higher sensitivity by true capillary design
  • The ultimate level of performance and versatility in GC-IRMS, including the hyphenation with a quadrupole MS
Thermo Scientific LC IsoLink II IRMS System
  • Space-saving design with intuitive system operation
  • Novel intelligence features of the Vanquish Core HPLC System
  • A unique backflush function for significant system uptime and productivity
Videos
Thermo Scientific Vanquish Fraction Collector
  • Innovative valve technology for enhanced resolution, product recovery, and minimal carry-over
  • Compatible with any analytical flow Vanquish HPLC and UHPLC System
  • Broad flow range from 0.05 – 10.0 ml/min for wide application flexibility
Equine sports testing
With an increase in anabolic androgenic steroids (e.g.: hemapolin and oxandrolone) in positive case reports, the most dominant analytical technique within equine sports testing remains LC-MS, which includes broad screening methods to include horsehair matrices. Additionally, while primarily focused on the human doping field, the role of peptides and macromolecules such as erythropoietin (EPO), recombinant human EOP (rhuEPO) and biomarker identification (e.g. testosterone esters) are equally applicable to equine doping. Finally, the use of ion chromatography mass spectrometry (IC-MS) has emerged as a more streamlined technique for identification of bisphosphonates, a compound generally used to manage bone issues in horses.
Equine sports testing workflow
Equine sport testing laboratories may run thousands of samples each year, putting their reputations on the line. The volume of samples, complex sample matrices, variety of compounds, and unique small molecules mean sample prep techniques must require minimal time while offering fast throughput. Thermo Fisher Scientific offers solutions to help maximize sample clean up and improve your analytical sensitivity.
Thermo Scientific SOLAμ SPE Plates
  • Reproducible, robust sample extraction using very low samples and solvent volumes
  • Award-winning innovative frit-less SPE technology
  • Low analytical sample failures
Applied Biosystems KingFisher Flex Purification System
  • Fully automated system yields high-speed purification of nucleic acids, proteins and cells
  • Customers can raise the processing volume up to 5 ml, for increased yields
  • Ideal for targeted identification, veterinary assays, and biomarker analysis
Multiple governing bodies control doping in racehorses. These include the Fédération Equestre Internationale (FEI), Horseracing Integrity and Safety Authority (HISA), United States Equestrian Federation (USEF), Horseracing Integrity & Welfare Unit (HIWU), to name a few.
As a result, antidoping regulations can vary, sometimes significantly, from one country to another. Additionally, there is no official standardization on thresholds, screening methods, screening limits, or what to do with the B sample—the “verification” sample and how to test it. Working with industry leaders, we continue to support leading edge equine testing.
Thermo Scientific Transcend TurboFlow System
Thermo Scientific Vanquish Flex with Thermo Scientific Altis Plus MS
Thermo Scientific Dionex ICS-6000 HPIC IC System
Thermo Scientific Orbitrap Exploris 240 Mass Spectrometer with Vanquish UHPLC System
  • Chromatography for online sample clean-up and improved LC-MS detection
  • Increase productivity with multi-channel LC capabilities
  • Thermo Scientific Aria MX SW operates each channel independently for single or multi-method operation
  • Quantitation for challenging molecules in simple to complex matrices
  • Up to 800 SRM/s increase the amount of molecular quantitation in less time
  • Selectivity with high-resolution (0.2 Da FWHM) for improved signal to noise ratios
  • Modular ion chromatography (IC) system capable of operation up to 5000 psi
  • Excellent for screening and confirmatory analysis of bisphosphonates
  • System can be extended to other polar molecules
  • Resolving power from (FWHM) 15,000 to 240,000 at m/z 200 with exceptional mass accuracy
  • Mass accuracy with Thermo Scientific EASY-IC (internal calibration) ion source.
  • Maximum compound coverage with fast scan rates
TraceFinder data acquisition software is widely used for the analysis of doping compounds, including steroids, offering streamlined data processing and high-throughput screening capabilities. It facilitates both targeted and untargeted detection by integrating multi-channel LC, single quadrupole, triple quadrupole and high-resolution LC-MS systems, as well as IC and GC systems.
Thermo Scientific TraceFinder Software
  • Complete screening and quantitation software
  • Driven by powerful, intuitive acquisition and data processing
  • Smart sample flagging, flexible data review, and custom report generation removes bottlenecks associated with data review
Thermo Scientific Compound Discoverer Software
  • For untargeted screening, identify unknowns faster with connection to online mzCloud 2.0 spectral libraries or local Thermo Scientific mzVault spectral libraries
  • Fully integrated suite of advanced tools for both known-parent and unknown data processing and interpretation
  • Streamlines compound identification, comparative analyses
  • Provides extensive filtering and data visualization capabilities
Thermo Scientific Mass Frontier Spectral Interpretation Software
  • Increase unknown ID throughput and confidence
  • Know your unknowns
  • From spectra to structure
  • Build, store, share and leverage your knowledge
Thermo Scientific Chromeleon Chromatography Data System (CDS) Software
  • Supports targeted quantitation and screening MS workflows
  • Compliance and data integrity tools as standard
  • Improved sharing and security of data without need for a separate server
Research-based analysis
Healthcare today has witnessed the rapid development of new therapeutic approaches for various health-related diseases and disorders and offers more efficient and patient-oriented treatment options. However, these new drugs and methods can sometimes be misused to enhance athletic performance, posing challenges for antidoping programs. Additionally, the enhanced analytical power and inevitable exposure of elite athletes to environmental factors, chemicals and drugs, necessitates dedicated research in order to distinguish between unintentional contamination and intentional doping crucial for analytical testing laboratories.
Thermo Scientific Orbitrap Exploris 240 Mass Spectrometer
Thermo Scientific Orbitrap Excedion Pro Mass Spectrometer
Thermo Scientific Orbitrap IQ-X Tribrid Mass Spectrometer
Thermo Scientific Orbitrap Ascend Editions Tribrid Mass Spectrometer
  • Resolve more target ions from matrix interferences in complex samples with resolution up to 240,000 (FWHM) at m/z 200
  • Achieve sub 1-ppm mass accuracy levels with Thermo Scientific EASY-IC internal calibration source and single mass calibration for full mass range
  • Ultra-high-field Orbitrap mass analyzer with scan speed up to 70 Hz
  • Improved sensitivity of low abundant compounds with enhanced dynamic range (eDR)
  • Lower limits of detection for a wider range of molecules
  • Designed for unknown compound ID and small molecule analysis
  • Combination of the best of quadrupole, linear ion trap and Orbitrap technology for acquisition of the richest MSn data
  • Optional 1,000,000 (1M) FWHM resolution at m/z 200 for improved unknown analysis
  • Native MS option for improved selectivity and MSn sensitivity of macromolecules
  • PTCR option to reduce m/z spectrum complexity
  • UVPD option for improved fragmentation
Thermo Scientific FAIMS Pro Duo Interface
FAIMS technology provides an additional dimension of separation to enhance LC-MS analysis. This results in improved signal-to-noise and LOQ when dealing with complex matrices, such as human serum.
Thermo Scientific Compound Discoverer Software
  • Fully integrated suite of advanced tools for both known-parent and unknown data processing and interpretation
  • Streamlines compound identification, comparative analyses
  • Provides extensive filtering and data visualization capabilities
Thermo Scientific Mass Frontier Spectral Interpretation Software
  • Increase unknown ID throughput and confidence
  • Know your unknowns
  • From spectra to structure
  • Build, store, share and leverage your knowledge
Thermo Scientific Proteome Discoverer Software
  • Unifying protein informatics platform – DIA-DDA-WWA
  • Simplifies the identification and quantification of proteins in complex biological samples
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.