Orbitrap™ Ascend Editions Tribrid™ Mass Spectrometer
Orbitrap™ Ascend Editions Tribrid™ Mass Spectrometer
Orbitrap™ Ascend Editions Tribrid™ Mass Spectrometer
Orbitrap™ Ascend Editions Tribrid™ Mass Spectrometer
Thermo Scientific™

Orbitrap™ Ascend Editions Tribrid™ Mass Spectrometer

Scale up your science for proteomics and native protein analysis with versatility and ease-of-use with Orbitrap Ascend Editions Tribrid mass spectrometers.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
FSN06-10000Orbitrap Ascend Tribrid MS
B51003847Orbitrap Ascend MultiOmics Tribrid MS
B51003848Orbitrap Ascend Structural Biology Tribrid MS
B51003849Orbitrap Ascend BioPharma Tribrid MS
Catalog number FSN06-10000
Price (MXN)
-
Request A Quote
Model:
Orbitrap Ascend Tribrid MS

Exceed experimental expectation and versatility with recommended MultiOmics, Structural Biology, and BioPharma configurations of the Thermo Scientific™ Orbitrap Ascend Editions Tribrid™ Mass Spectrometer. Each Edition provides a tailored configuration of options for each application space, simplifying your decisions and streamlining your research. Characterize even the largest biopharmaceuticals and protein complexes with the Native MS option, which extends the Orbitrap mass range up to m/z 16,000 and new quadrupole isolation up to m/z 8,000. Unlock the potential of alternate fragmentation techniques to decipher the most challenging molecules.

With three unique fit-for-purpose Editions that are tailored for MultiOmics; Structural Biology; and BioPharma application areas, Orbitrap Ascend Editions Tribrid mass spectrometers ensure high-quality data acquisition. Whether you're focused on multiplexed quantitative proteomics, metabolomics, lipidomics or glycomics, small molecule-based pharmaceuticals or biotherapeutics, single-cell or deciphering higher-order protein structures; these instruments are uniquely suited to your needs.

  • Native MS option: Orbitrap extended mass range up to m/z 16000 and new quadrupole isolation up to m/z 8000 for improved selectivity and MSn sensitivity of macromolecules.
  • PTCR option: Reduce m/z spectrum complexity to enhance proteoform characterization at MS1 and MS2 levels.
  • UVPD option: Improved fragmentation for top-down proteomics, oligonucleotides, lipidomics, and other analytes
  • Easy-ETD option: Achieve high sequence coverage of proteins, including site localization of labile PTMs.
  • EASY-IC ion source option: Obtain sub-1ppm mass accuracy with internal calibration.
  • Dual IRMs: Confidently identify more molecules with two ion-routing multiples to increase scan rate and sensitivity.
  • Real-Time Library Search: Provides higher confidence identifications for peptide or small molecule analysis using MSn.
  • Auto-Ready ion source: Spend less time on instrument setup with a new dedicated source for autocalibration.
  • Pre-built method templates: Optimized methods for each Edition system, covering diverse applications.
  • Dry-Pump: Environmentally friendly, oil-free dry pump is standard to decrease maintenance while providing leak-free vacuum performance.
Specifications
Dimensions (D x W x H)32 x 50 x 28 in. (81 x 127 x 70 cm)
Dynamic Range> 5000 within a single scan
Mass Accuracy< 3 ppm RMS using external calibration
ModelOrbitrap Ascend Tribrid MS
Polarity Switching

One full experimental cycle acquired in <1.1 seconds

Scan RatesUp to 45 Hz
Weight (Metric)181 kg (400 lb)
Mass Range40 to 6000 m/z
Max. ResolutionUp to 480,000 at m/z 200
TypeTribrid MS, Standard
Unit SizeEach