TSQ Altis™ Plus Triple Quadrupole Mass Spectrometer
TSQ Altis™ Plus Triple Quadrupole Mass Spectrometer
TSQ Altis™ Plus Triple Quadrupole Mass Spectrometer
TSQ Altis™ Plus Triple Quadrupole Mass Spectrometer
TSQ Altis™ Plus Triple Quadrupole Mass Spectrometer
Thermo Scientific™
TSQ Altis™ Plus Triple Quadrupole Mass Spectrometer
Leverage innovative hardware and software advancements to take sensitivity, selectivity, and data-acquisition performance for targeted analyses to the ultimate level.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMass RangeType
TSQ03-100022 to 2010 m/zTSQ LC-MS with Oil Pump
TSQ03-110022 to 2010 m/zTSQ LC-MS with Dry Pump
Catalog number TSQ03-10002
Price (USD)
-
Mass Range:
2 to 2010 m/z
Type:
TSQ LC-MS with Oil Pump
Overcome the most demanding quantitative workflow challenges with the TSQ Altis™ Plus triple quadrupole mass spectrometer. With superior acquisition speeds, enhanced sensitivity, prototypical selectivity, and exceptional robustness, the TSQ Altis Plus mass spectrometer delivers unprecedented accuracy and precision for low-level compound detection and quantitation in complex matrices that redefine ultimate instrument performance.
The TSQ Altis Plus triple quadrupole mass spectrometer, together with industry-leading quantitation software, UHPLC and HPIC systems, and superior sample preparation kits; helps researchers overcome complex analytical challenges using robust, reliable, and sensitive targeted quantitation. The main benefits of the TSQ Altis Plus mass spectrometer include:
  • Ultimate sensitivity for all molecular types in matrices ranging from simple to complex
  • Outstanding instrument robustness, enabling increased confidence in generated data with no loss of instrument uptime
  • Ultrafast selected-reaction monitoring (SRM), increasing the amount of molecular quantification in less time
  • Exceptional selectivity with high-resolution (0.2 Da FWHM) selected-reaction monitoring (H-SRM) performance to improve signal-to-noise ratios for targeted compound
  • Close integration with application-specific software, increasing productivity across all markets and applications
  • Simplicity and ease-of-use, allowing users of all expertise levels to acquire high-quality results with increased confidence
Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.
For General Lab Use Only. Not For Diagnostic Procedures.
Specifications
For Use With (Application)Pharmaceutical, Environmental, Food Safety, Clinical Research, Forensic Toxicology
For Use With (Equipment)OptaMax NG ion source, FAIMS Pro Duo interface, VeriSpray PaperSpray Ion Series ion source, Nanospray Flex Series ion source
Mass Range2 to 2010 m/z
TypeTSQ LC-MS with Oil Pump
Unit SizeEach