Search
Light weight and shatterproof, Thermo Scientific Nalgene vacuum chambers and desiccators provide a safer, more ergonomic alternative to glass. These products can make significant contributions to your lab safety program. Make your vacuum connections with non-toxic Nalgene orthophthalate-free vacuum tubing. For field applications and areas with limited access to an electric-powered vacuum source, Nalgene hand-operated vacuum pumps or a water-generated vacuum provided by a Nalgene vacuum aspirator may provide a low cost, convenient solution.
Plastic vacuum chambers can play an important role in lab safety programs. They don't shatter upon impact like glass and are much lighter weight for more ergonomic handling.
Nalgene Vacuum Chambers | Nalgene Replacement Vacuum Jars | Nalgene Replacement Plate for Vacuum Chambers |
![]() | ![]() | ![]() |
| Recommended as a controlled environment for degassing operations and freeze drying | Replacement jars for Nalgene vacuum chambers | Replacement base plate for Nalgene vacuum chambers |
| Order | Order | Order |
Light weight and shatterproof, these desiccators are an excellent choice for improved lab safety.
Nalgene Transparent PC Classic Design Desiccator | Nalgene Transparent PC Classic Design Desiccator |
![]() | ![]() |
| Classic shape | Space ship body design |
| Red rubber gasket seal | No gasket |
| No grease needed | Stopcock grease seal |
| Larger volume | Lower profile |
| Order | Order |
Nalgene Non-Phthalate Clear Plastic PVC Vacuum Tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene 180 tubing but is free from orthophthalate DEHP plasticizers. Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and complies with California proposition 65. Resists hardening and discoloration from a wide range of laboratory chemicals and does not impart odor or taste. Withstands full vacuum (29.9 in. Hg) at room temperature. Use 1/4" ID sizes with Nalgene vacuum chambers, hand pumps, vacuum desiccators and vacuum aspirator.
Hand-operated vacuum pumps are a convenient choice in the lab or field where an electric-generated vacuum source is unavailable. Repair kits are available to keep them operating at top efficiency for extended product life and sustainable "fix-it-don't replace-it" value support. Pumps feature full blank port pressure with only two strokes and single finger trigger release for easy and controlled rate of vacuum release. Nozzle fits standard 1/4" (6 mm) ID tubing and each unit comes with a 2 ft. (0.6 m) length of Nalgene PVC vacuum tubing.
Zinc/Aluminum Vacuum Pump with gauge | PVC Vacuum Pump with gauge, 36 cc | PVC Vacuum Pump with gauge, 15 cc | PVC Vacuum Pump without gauge, 36 cc | PVC Vacuum Pump without gauge, 15 cc |
![]() | ![]() | ![]() | ![]() | ![]() |
| Durable hand pump recommended for small jobs around the lab or in the field where an electric pump is not available | Recommended for small volume filtration jobs, siphoning, liquid transfer, bleeding fluid lines and checking for leaks | Ideal for siphoning, liquid transfer, bleeding fluid lines and checking for leaks | Economy version suitable for small volume filtration jobs, siphoning, liquid transfer, bleeding fluid lines and checking for leaks | Economy version good for field or lab use |
| Zn/Al Metal body and hand grip | PVC body and hand grip | |||
| Guage included | No gauge | |||
| 15 cc per stroke | 36 cc per stroke | 15 cc per stroke | 36 cc per stroke | 15 cc per stroke |
| Max. pressure 30psi (207 Kpa) | No pressure capability | |||
| Max. Vacuum 25 in. (635mm) Hg | ||||
| Repair Kit available # 6133-2000 | Repair Kit available # 6132-3000 | Repair Kit available # 6132-2000 | Repair Kit available # 6132-3000 | Repair Kit available # 6132-2000 |
| Order | Order | Order | Order | Order |
Attach the Nalgene Aspirator Vacuum Pump directly to sink faucets for vacuum draw. It can draw 11.5L air per minute at a water flow rate of 6.5L per minute. It operates efficiently with water pressures as low as 7.5 psig for minimum water consumption and comes with an integral check valve and tubing for insertion in discharge end.
Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.