Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 4 | 750 | Cy7 | 757 | 783 | 5 (in antifade) | Microscopy |
Invitrogen Aluora dyes, including Aluora 750 dye, are a specialized organic dye technology specifically optimized for spatial biology and multiplex immunohistochemistry (IHC) applications. Aluora 750 dye is excited by the 750 nm laser and emits at 783 nm. This dye has exceptional brightness and photostability. To learn more about spatial imaging, visit our Spatial Biology Resource Center.
We offer Aluora 750 spatial amplification reagents for use in immunohistochemistry.
Human FFPE small intestine was incubated with 1:1,000 of Biotinylated Ki-67 Monoclonal Antibody (SolA15) (Cat. No. 13-5698-82) then labeled with the reagents in the Aluora 750 HRP-streptavidin Kit (Cat. No. A40001352) and a DAPI nuclear stain. The slides were mounted in ProLong Glass mountant. Images were captured with an Invitrogen EVOS M7000 instrument (see information about EVOS imaging systems).
Invitrogen Aluora dyes are a specialized organic dye technology specifically optimized for spatial biology and multiplex immunohistochemistry applications. These bright, photostable dyes are excellent for detecting extremely low-abundance targets. Additionally, Aluora dyes exhibit a distinct emission spectra fingerprint that can be spectrally unmixed for high plex imaging experiments on spatial imaging platforms such as the EVOS S1000 Spatial Imaging System.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.