Search
Peridinin chlorophyll protein-Cyanine5.5 (PerCP-Cyanine5.5) is a photostable tandem dye excited from the 488 nm laser. It is stable in fixative, and stained cells can be stored overnight. Less-abundant intracellular antigens stained with PerCP-Cyanine5.5 can often be reliably detected in multicolor flow cytometry experiments.
We offer PerCP-Cyanine5.5 dye conjugated to a variety of antibodies.
Initial brightness | PerCP-Cyanine5.5 dye is a near-IR fluorophore used primarily for flow cytometry, optimally excited by the blue laser. | ||
Photostability in buffer |
| Medium | 488 | 695/40 | 482 | 690 | ||
Relative staining index | Laser line | Common filter set | Excitation max | Emission max |
We offer PerCp-Cyanine5.5 dye conjugated to a variety of antibodies and streptavidin conjugates.