eBioscience™ Streptavidin PerCP-Cyanine5.5 Conjugate
Invitrogen™

eBioscience™ Streptavidin PerCP-Cyanine5.5 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
45-4317-82100 μg
45-4317-8025 μg
Catalog number 45-4317-82
Price (USD)
276.65
Online exclusive
369.00
Save 92.35
Each
Quantity:
100 μg
Price (USD)
276.65
Online exclusive
369.00
Save 92.35
Each

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Blue Laser

Emit

695 nm

Excite

488 nm

Reported Application

Flow Cytometric Analysis

For Research Use Only
Specifications
Concentration0.2 mg/mL
Label or DyePerCP-Cy5.5
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity100 μg
Target MoleculeBiotin
ConjugatePerCP-Cy5.5
FormLiquid
Product LineeBioscience
Unit SizeEach