Search
They won’t break, chip or shatter during use and cleaning. Lightweight and easy to handle. Chemical resistance, heat tolerance (not for use on hot plates), and spout design make Thermo Scientific Nalgene Griffin low-form beakers a perfect long-term solution for your graduated beaker choice. Meeting the ISO/DIN 7056 International Standards for Laboratory Plastic Beakers, our polypropylene and polymethylpentene beakers enable precise measurements. Nalgene beakers with handles offer reliable support and dispensing control. Nalgene beakers are made only from high quality laboratory grade plastic materials for dependably lower levels of potential extractables and leachables (E&L), so you can rely on them for everyday lab use without interfering with your work.
Looking for ways to make your lab more sustainable? Nalgene polypropylene copolymer beakers are reusable, durable and recyclable at end-of-life in many communities; discover more about the sustainability of using reusable plastic labware and Thermo Fisher Scientific's green manufacturing practices through our participation in My Green Lab's ACT Label program.
| Nalgene PPCO Griffin Low-Form Plastic Beakers | Nalgene PMP Griffin Low-Form Plastic Beakers | Nalgene HDPE Graduated Plastic Beakers with Handles | Nalgene PMP Graduated Plastic Beakers with Handles | Nalgene PFA Graduated Plastic Beakers with Handles | |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| Milky white, graduated, good chemical resistance | Transparency of glass, graduated, excellent chemical resistance, heat tolerance | Milky white, heavy wall, ultimate chemical resistance, heat tolerance | Milky white, extra-heavy wall, sturdy rectangular handle, reliable support, dispensing control | Transparent, graduated, sturdy rectangular handle, easy, safe handling and pouring | Milky white, heavy wall, ultimate chemical resistance, heat tolerance |
| Meets ISO/DIN 7056**, 21 CFR Pt 177.1520 | Meets ISO/DIN 7056"", 21 CFR Pt 177.1520 | — | Meets 21 CFR Pt 177.1520 | Meets 21 CFR Pt 177.1520 | — |
| Autoclavable | Autoclavable | Autoclavable | Not autoclavable | Autoclavable | Autoclavable |
| Max use 121oC* | Continuous use 150oC intermittent use up to 170oC* | Max Use 250oC* | Max use 120oC* | Continuous use 150oC intermittent use up to 170oC* | Max Use 250oC* |
| Graduations: silk screened | Graduations: silk screened | No graduations | Graduations: molded-in; oz and mL | Graduations: molded-in; oz and mL | Graduations: molded-in; oz and mL |
| Volume range: 30mL–4L | Volume range: 50mL–4L | Volume range: 50mL–1L | Volume range: 1L, 3L | Volume range: 1L, 3L | Volume range: 1L, 3L |
| PPCO | PMP | PFA | HDPE | PMP | PFA |
| Translucent white | Clear | Translucent white | Translucent white | Clear | Translucent white |
| No handle | No handle | No handle | Handle | Handle | Handle |
| Nesting | Nesting | Nesting | Not nesting | Not nesting | Not nesting |
| BPA free | BPA free | BPA free | BPA free | BPA free | BPA free |
| Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
| ACT Label › | |||||
| Order Now | Order Now | Order Now | Order Now | Order Now | Order Now |
*Nalgene beakers are not for use on hot plates.
**International Standards for Laboratory Plasticware
| Nalgene PPCO Griffin Low-Form Plastic Beakers | Nalgene PMP Griffin Low-Form Plastic Beakers | Nalgene Graduated HDPE Plastic Beakers with Handles | Nalgene PMP Graduated Plastic Beakers with Handles | Nalgene PFA Graduated Plastic Beakers with Handles | ||
| Acids, dilute or weak | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Acids^ strong, concentrated | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Alcohols, aliphatic | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Aldehydes | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Bases/alkali | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Esters | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Hydrocarbons, aromatic | — | — | ✓ | — | — | ✓ |
| Hydrocarbons, halogenated | — | — | ✓ | — | — | ✓ |
| Ketones, aromatic | — | ✓ | ✓ | — | ✓ | ✓ |
| Oxidizing agents, strong | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| ✓ | ✓ | ✓ | ✓ | ✓ |
| ✓ | ✓ | ✓ | ✓ | ✓ |
+ Ratings are for short term intermittent exposure. Wash thoroughly between uses.
^ Except oxidizing acids; for oxidizing acids, see “Oxidizing agents, strong.”
Thermo Scientific Nalgene Economy Beakers are a simplified version of the original Nalgene premium beakers for those with more basic product requirements. The Economy Beakers are made from high quality laboratory grade polypropylene copolymer for reliably low extractables and shatter-proof performance at temperatures above -40°C, just like Nalgene premium beakers. The molded in-nominal graduations found on the Economy Beakers differ from the premium beaker’s high visibility printed graduations and, thus, reduces cost
Your work depends on quality and consistency, and we believe every lab should have the best tools. Watch Video
When looking for sustainable products, we believe it should be easy to make informed purchasing decisions. We're working to provide more information about our products' environmental impact by participating in the ACT Label program with a selection of Thermo Scientific Nalgene and Matrix products. It's like an eco-nutrition label for sustainability.
Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.