Dr. Koefoed-Nielsen presents data for a patient who had undergone two kidney transplantations, experienced allograft failure soon after receiving a third kidney transplant. With a negative crossmatch and no DSA detected, further investigation was required. Non-HLA autoantibodies can be associated with adverse transplantation outcomes1, so they screened the patient and eight others who experienced similar outcomes for autoantibodies using the One Lambda™ LABScreen™ Autoantibody Group 1 and Group 2 assays. All nine patients had high levels of REG3A, LMNB, PRKCH, GAPDH, HNRNPK, VM, and IFNG autoantibodies, which has been correlated with humoral rejection in kidney and heart transplant recipients.
Speaker:
Pernille Koefoed-Nielsen, MD, PhD
Tissue Typing Laboratory,
Department of Clinical Immunology
Aarhus Universitet Hospital, Denmark
Recorded:
Nov 24, 2021
Support
Please sign in to access your account. Customers must log in to access some content, recorded webinars, training materials, documents, and software.
Please provide the following to setup your user account. If you do not know your center ID, please leave the center ID field blank.
Passwords do not match