Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HTRA2
Transcript:
NM_013247.4
Assay Name:
Mutation genome location:
chr.2 74531771-74532274 on build GRCh38
Gene:
LOXL3
Transcript:
NM_013247.4
Assay Name:
Mutation genome location:
chr.2 74531771-74532274 on build GRCh38
Assay ID
Hs00195707_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
serine protease
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
ceramide metabolic process
mitochondrion organization
adult walking behavior
intrinsic apoptotic signaling pathway in response to DNA damage
response to herbicide
positive regulation of cell death
protein autoprocessing
pentacyclic triterpenoid metabolic process
forebrain development
cellular response to oxidative stress
cellular response to heat
cellular response to interferon-beta
regulation of multicellular organism growth
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
cellular protein catabolic process
negative regulation of cell cycle
neuron development
cellular response to retinoic acid
cellular response to growth factor stimulus
execution phase of apoptosis
negative regulation of neuron death
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
epithelial to mesenchymal transition
receptor-mediated endocytosis
negative regulation of transcription, DNA-templated
oxidation-reduction process
serine-type endopeptidase activity
protein binding
peptidase activity
serine-type peptidase activity
unfolded protein binding
protein-lysine 6-oxidase activity
scavenger receptor activity
copper ion binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline