Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TLR9
Transcript:
NM_017442.3
Assay Name:
Mutation genome location:
chr.3 52221774-52222278 on build GRCh38
Assay transcription exon location:
Within Exon 2
Assay ID
Hs00233007_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
toll-like receptor signaling pathway
response to molecule of bacterial origin
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
I-kappaB phosphorylation
axonogenesis
positive regulation of gene expression
maintenance of gastrointestinal epithelium
negative regulation of NF-kappaB transcription factor activity
tumor necrosis factor production
negative regulation of interleukin-6 production
negative regulation of interleukin-8 production
positive regulation of chemokine production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-beta production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-18 production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
negative regulation of toll-like receptor signaling pathway
positive regulation of toll-like receptor signaling pathway
toll-like receptor 9 signaling pathway
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of NF-kappaB import into nucleus
defense response to bacterium
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
positive regulation of interferon-gamma biosynthetic process
innate immune response
positive regulation of interferon-alpha biosynthetic process
positive regulation of interferon-beta biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
regulation of cytokine secretion
positive regulation of inflammatory response
defense response to Gram-negative bacterium
positive regulation of NF-kappaB transcription factor activity
positive regulation of nitric-oxide synthase biosynthetic process
negative regulation of calcium-transporting ATPase activity
transmembrane signaling receptor activity
interleukin-1 receptor binding
1-phosphatidylinositol-3-kinase activity
siRNA binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-vps2j:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0