Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00443812_cn
          See other SBNO2 CNV Assays ›
          Gene Symbol
          SBNO2
          Assay Reference Genome
          Location

          Chr.19:1111589 on build GRCh38
          Cytoband
          19p13.3
          Assay ID Hs00443812_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          22904

          Gene Name:

          strawberry notch homolog 2

          Gene Aliases:

          KIAA0963, SNO, STNO

          Location:

          Chr.19:1107634-1174283 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 24 - Exon 25
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SBNO2 NM_001100122.1 NP_001093592.1
          NM_014963.2 NP_055778.2
          XM_011527804.2 XP_011526106.1
          AB023180.1
          AK074102.1 BAB84928.1
          AK125139.1
          AK292407.1
          AK296849.1
          AK300981.1
          BC106021.1 AAI06022.1
          BC127011.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1058677 Chr.19:1054061 - 1283189 on Build GRCh38 Loss ARHGAP45 CBARP MIDN CIRBP-AS1 STK11 GPX4 SBNO2 EFNA2 C19orf24 LOC102725180 CIRBP POLR2E ATP5D ABCA7
          nsv1056200 Chr.19:1107036 - 1193347 on Build GRCh38 Gain SBNO2 LOC102725180
          dgv136n111 Chr.19:1038223 - 1329640 on Build GRCh38 Deletion ARHGAP45 CBARP CNN2 MIDN CIRBP-AS1 STK11 GPX4 SBNO2 EFNA2 C19orf24 LOC102725180 CIRBP POLR2E ATP5D ABCA7
          nsv470102 Chr.19:1046521 - 1152656 on Build GRCh38 Loss ARHGAP45 SBNO2 POLR2E ABCA7 GPX4
          nsv516646 Chr.19:1110830 - 1146101 on Build GRCh38 Loss SBNO2
          nsv470103 Chr.19:1086044 - 1325151 on Build GRCh38 Gain ARHGAP45 CBARP MIDN CIRBP-AS1 STK11 GPX4 SBNO2 EFNA2 C19orf24 LOC102725180 CIRBP POLR2E ATP5D
          nsv953939 Chr.19:1104902 - 1174301 on Build GRCh38 Deletion SBNO2 LOC102725180 GPX4
          nsv9662 Chr.19:893394 - 1317740 on Build GRCh38 Gain ARHGAP45 CBARP R3HDM4 CNN2 RNU6-9 KISS1R GRIN3B MIDN CIRBP-AS1 RNU6-2 STK11 TMEM259 GPX4 SBNO2 EFNA2 LOC105372235 C19orf24 LOC102725180 CIRBP POLR2E ARID3A WDR18 ATP5D ABCA7
          nsv1160555 Chr.19:887275 - 1346084 on Build GRCh38 Deletion ARHGAP45 CNN2 RNU6-9 KISS1R GRIN3B MIDN CIRBP-AS1 RNU6-2 TMEM259 SBNO2 LOC105372235 LOC102725180 CIRBP ATP5D CBARP R3HDM4 STK11 GPX4 EFNA2 C19orf24 MED16 POLR2E ARID3A WDR18 ABCA7
          nsv469845 Chr.19:951640 - 1144486 on Build GRCh38 Loss ARHGAP45 SBNO2 LOC105372235 CNN2 GRIN3B POLR2E RNU6-2 ARID3A WDR18 ABCA7 TMEM259 GPX4
          nsv833704 Chr.19:951641 - 1144486 on Build GRCh38 Gain ARHGAP45 SBNO2 LOC105372235 CNN2 GRIN3B POLR2E RNU6-2 ARID3A WDR18 ABCA7 TMEM259 GPX4
          nsv578215 Chr.19:1095062 - 1124032 on Build GRCh38 Gain SBNO2 POLR2E GPX4
          dgv135n111 Chr.19:1014378 - 1156398 on Build GRCh38 Deletion ARHGAP45 SBNO2 CNN2 POLR2E RNU6-2 ABCA7 TMEM259 GPX4
          nsv828367 Chr.19:1102094 - 1113817 on Build GRCh38 Gain SBNO2 GPX4
          nsv1071487 Chr.19:1111500 - 1112601 on Build GRCh38 Deletion SBNO2

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs78026836] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s)

          macrophage activation involved in immune response
          transcription, DNA-templated
          bone mineralization
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of inflammatory response
          bone trabecula morphogenesis
          cellular response to lipopolysaccharide
          cellular response to interleukin-11
          cellular response to interleukin-6
          osteoclast fusion
          cellular response to leukemia inhibitory factor

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline