Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00556119_cn
          See other TCF7L2 CNV Assays ›
          Gene Symbol
          TCF7L2
          Assay Reference Genome
          Location

          Chr.10:112951233 on build GRCh38
          Cytoband
          10q25.2
          Assay ID Hs00556119_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6934

          Gene Name:

          transcription factor 7 like 2

          Gene Aliases:

          TCF-4, TCF4

          Location:

          Chr.10:112950219-113167678 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 1 - Exon 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TCF7L2 NM_001146274.1 NP_001139746.1
          NM_001146283.1 NP_001139755.1
          NM_001146284.1 NP_001139756.1
          NM_001146285.1 NP_001139757.1
          NM_001146286.1 NP_001139758.1
          NM_001198525.1 NP_001185454.1
          NM_001198526.1 NP_001185455.1
          NM_001198527.1 NP_001185456.1
          NM_001198528.1 NP_001185457.1
          NM_001198529.1 NP_001185458.1
          NM_001198530.1 NP_001185459.1
          NM_001198531.1 NP_001185460.1
          NM_030756.4 NP_110383.2
          XM_005270071.1 XP_005270128.1
          XM_005270075.1 XP_005270132.1
          XM_005270077.1 XP_005270134.1
          XM_005270078.1 XP_005270135.1
          XM_005270079.1 XP_005270136.1
          XM_005270080.1 XP_005270137.1
          XM_005270084.1 XP_005270141.1
          XM_005270085.1 XP_005270142.1
          XM_005270086.1 XP_005270143.1
          XM_005270088.1 XP_005270145.1
          XM_005270089.1 XP_005270146.1
          XM_005270091.2 XP_005270148.1
          XM_005270092.1 XP_005270149.1
          XM_005270093.2 XP_005270150.1
          XM_005270094.2 XP_005270151.1
          XM_005270095.1 XP_005270152.1
          XM_005270096.2 XP_005270153.1
          XM_005270100.1 XP_005270157.1
          XM_005270101.2 XP_005270158.1
          XM_005270102.1 XP_005270159.1
          XM_005270103.1 XP_005270160.1
          XM_005270104.1 XP_005270161.1
          XM_011540109.1 XP_011538411.1
          XM_011540110.1 XP_011538412.1
          XM_011540111.1 XP_011538413.1
          XM_011540113.2 XP_011538415.1
          XM_011540116.1 XP_011538418.1
          XM_017016584.1 XP_016872073.1
          XM_017016585.1 XP_016872074.1
          XM_017016586.1 XP_016872075.1
          XM_017016587.1 XP_016872076.1
          XM_017016588.1 XP_016872077.1
          XM_017016589.1 XP_016872078.1
          XM_017016590.1 XP_016872079.1
          XM_017016591.1 XP_016872080.1
          XM_017016592.1 XP_016872081.1
          XM_017016593.1 XP_016872082.1
          XM_017016594.1 XP_016872083.1
          XM_017016595.1 XP_016872084.1
          XM_017016596.1 XP_016872085.1
          AB440195.1
          AB451266.1
          AK225809.1
          AK299295.1
          AK315243.1
          BC032656.1 AAH32656.1
          BX648364.1
          CR536574.1 CAG38811.1
          FJ010164.1 1 111
          FJ010165.1 1 111
          FJ010166.1
          FJ010167.1
          FJ010168.1
          FJ010169.1
          FJ010170.1
          FJ010171.1
          FJ010172.1
          FJ010173.1 1 111
          FJ010174.1
          HM352838.1
          HM352839.1
          HM352840.1
          HM352841.1
          HM352842.1
          HM352843.1
          HM352844.1
          HM352845.1
          HM352846.1
          HM352847.1
          HM352848.1
          HM352849.1
          HM352850.1
          HM352851.1
          Y11306.2 CAA72166.2

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv518n106 Chr.10:112949441 - 112951941 on Build GRCh38 Deletion TCF7L2
          nsv1070891 Chr.10:112949140 - 112951941 on Build GRCh38 Deletion TCF7L2

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          cell cycle arrest
          Wnt signaling pathway, calcium modulating pathway
          cell proliferation
          response to glucose
          positive regulation of heparan sulfate proteoglycan biosynthetic process
          pancreas development
          positive regulation of insulin secretion
          positive regulation of protein binding
          regulation of hormone metabolic process
          glucose homeostasis
          negative regulation of sequence-specific DNA binding transcription factor activity
          maintenance of DNA repeat elements
          canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
          fat cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of protein export from nucleus
          myoblast fate commitment
          regulation of smooth muscle cell proliferation
          positive regulation of protein kinase B signaling
          canonical Wnt signaling pathway
          negative regulation of canonical Wnt signaling pathway
          beta-catenin-TCF complex assembly
          negative regulation of type B pancreatic cell apoptotic process
          negative regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II repressing transcription factor binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          beta-catenin binding
          transcription factor binding
          protein kinase binding
          nuclear hormone receptor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          gamma-catenin binding
          armadillo repeat domain binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline