Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01277368_cn
          See other PDE8A CNV Assays ›
          Gene Symbol
          PDE8A
          Assay Reference Genome
          Location

          Chr.15:85120885 on build GRCh38
          Cytoband
          15q25.3
          Assay ID Hs01277368_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5151

          Gene Name:

          phosphodiesterase 8A

          Gene Aliases:

          HsT19550

          Location:

          Chr.15:84980513-85139145 on Build GRCh38

          Assay Gene Location:

          Within Exon 22
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PDE8A NM_001243137.1 18 1810 NP_001230066.1
          NM_002605.2 18 2012 NP_002596.1
          NM_173454.1 17 1874 NP_775656.1
          XM_011521668.2 11 2146 XP_011519970.1
          XM_017022308.1 15 2243 XP_016877797.1
          XM_017022309.1 16 1610 XP_016877798.1
          XM_017022311.1 19 2747 XP_016877800.1
          AF056490.1 16 1475 AAC39763.1
          AF332653.1 18 1959 AAK57641.1
          AF388183.1 18 2005 AAL18610.1
          AF388184.1 17 1867 AAL18611.1
          AF388185.1 16 1788 AAL18612.1
          AF388186.1 17 1926 AAL18613.1
          AF388187.1 19 2203 AAL18614.1
          AK074280.1 18 1810
          AK122689.1 1 695
          AK127232.1 15 3293
          BC048317.1 2 252 AAH48317.1
          BC060762.1 19 2075 AAH60762.1
          BC075822.1 18 1903 AAH75822.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv94721 Chr.15:85119482 - 85128052 on Build GRCh38 Deletion PDE8A
          nsv1646 Chr.15:85089181 - 85122589 on Build GRCh38 Insertion PDE8A
          esv3581796 Chr.15:84996468 - 85167840 on Build GRCh38 Loss PDE8A
          nsv1043218 Chr.15:85092659 - 85183483 on Build GRCh38 Gain PDE8A
          nsv833078 Chr.15:85007383 - 85164833 on Build GRCh38 Loss PDE8A
          nsv833079 Chr.15:85076175 - 85274311 on Build GRCh38 Gain GOLGA6L3 LOC642423 LOC101929479 PDE8A
          nsv1037932 Chr.15:85118039 - 85168070 on Build GRCh38 Gain PDE8A
          nsv1048509 Chr.15:85020724 - 85175378 on Build GRCh38 Gain PDE8A
          esv2672879 Chr.15:84183834 - 85188520 on Build GRCh38 Deletion SCAND2P LINC00933 ZSCAN2 LOC105376722 GOLGA2P7 CSPG4P5 LOC103171574 NMB WDR73 GOLGA6L4 PDE8A UBE2Q2P1 LOC440300 SLC28A1 SEC11A GOLGA6L5P ZNF592 UBE2Q2L LOC102724135 ALPK3
          nsv428311 Chr.15:85107707 - 85486274 on Build GRCh38 Gain GOLGA6L3 LOC642423 MIR7706 AKAP13 LOC101929479 PDE8A

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          cAMP catabolic process
          regulation of transcription, DNA-templated
          signal transduction
          cyclic nucleotide metabolic process
          3',5'-cyclic-nucleotide phosphodiesterase activity
          3',5'-cyclic-AMP phosphodiesterase activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline