Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01622325_cn
          See other PAK6 CNV Assays ›
          Gene Symbol
          PAK6
          Assay Reference Genome
          Location

          Chr.15:40276359 on build GRCh38
          Cytoband
          15q15.1
          Assay ID Hs01622325_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          56924

          Gene Name:

          p21 (RAC1) activated kinase 6

          Gene Aliases:

          PAK5

          Location:

          Chr.15:40239091-40277487 on Build GRCh38

          Assay Gene Location:

          Within Exon 11
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PAK6 NM_001276717.1 NP_001263646.1
          NM_001276718.1 10 3088 NP_001263647.1
          NM_020168.5 11 2810 NP_064553.1
          AF276893.1 10 2509 AAF82800.1
          AJ236915.1 11 2750 CAC18720.1
          AK131522.1 10 3088
          BC035596.1 AAH35596.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv471240 Chr.15:40235954 - 40303426 on Build GRCh38 Loss PLCB2 C15orf56 PAK6 ANKRD63 BUB1B-PAK6
          nsv510660 Chr.15:40260705 - 40429684 on Build GRCh38 Deletion PLCB2 C15orf52 PHGR1 PAK6 KNSTRN INAFM2 DISP2 IVD ANKRD63 BUB1B-PAK6
          nsv569227 Chr.15:40273504 - 40314283 on Build GRCh38 Loss PLCB2 PAK6 ANKRD63 BUB1B-PAK6
          esv34145 Chr.15:40257416 - 40597626 on Build GRCh38 Loss C15orf57 CHST14 PHGR1 PAK6 DISP2 RPUSD2 MRPL42P5 ANKRD63 PLCB2 BAHD1 C15orf52 KNSTRN INAFM2 IVD KNL1 BUB1B-PAK6
          nsv517541 Chr.15:40265067 - 40303426 on Build GRCh38 Loss PLCB2 PAK6 ANKRD63 BUB1B-PAK6
          nsv952611 Chr.15:40237800 - 40284699 on Build GRCh38 Deletion C15orf56 PAK6 ANKRD63 BUB1B-PAK6

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          regulation of transcription, DNA-templated
          apoptotic process
          cytoskeleton organization
          signal transduction by protein phosphorylation
          cell-cell adhesion
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline