Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01663767_cn
          See other BEND7 CNV Assays ›
          Gene Symbol
          BEND7
          Assay Reference Genome
          Location

          Chr.10:13499835 on build GRCh38
          Cytoband
          Assay ID Hs01663767_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          222389

          Gene Name:

          BEN domain containing 7

          Gene Aliases:

          C10orf30

          Location:

          Chr.10:13438481-13529634 on Build GRCh38

          Assay Gene Location:

          Within Exon 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          BEND7 NM_001100912.2 2 979 NP_001094382.1
          NM_001369863.1 3 872 NP_001356792.1
          NM_001370075.2 3 872 NP_001357004.2
          NM_001378149.1 3 872 NP_001365078.1
          NM_001378150.1 3 872 NP_001365079.1
          NM_001378151.1 3 872 NP_001365080.1
          NM_001387359.1 3 872 NP_001374288.1
          NM_152751.3 3 1149 NP_689964.2
          XM_005252402.6 3 872 XP_005252459.1
          XM_011519390.3 3 702 XP_011517692.1
          XM_011519391.3 3 872 XP_011517693.2
          XM_011519392.3 3 872 XP_011517694.1
          XM_011519393.4 2 788 XP_011517695.1
          XM_011519395.3 2 979 XP_011517697.1
          XM_011519396.3 3 1070 XP_011517698.1
          XM_011519398.3 3 872 XP_011517700.2
          XM_011519399.3 3 872 XP_011517701.2
          XM_011519400.3 3 872 XP_011517702.2
          XM_011519402.3 3 872 XP_011517704.2
          XM_011519403.3 3 872 XP_011517705.2
          XM_011519404.4 3 872 XP_011517706.2
          XM_011519407.2 3 872 XP_011517709.2
          XM_011519411.3 3 872 XP_011517713.2
          XM_011519412.2 3 872 XP_011517714.2
          XM_047424785.1 3 872 XP_047280741.1
          XM_047424786.1 3 533 XP_047280742.1
          XM_047424787.1 3 414 XP_047280743.1
          XM_047424788.1 3 688 XP_047280744.1
          XM_047424789.1 4 808 XP_047280745.1
          XM_047424790.1 4 793 XP_047280746.1
          XM_047424792.1 4 978 XP_047280748.1
          XM_047424793.1 2 270 XP_047280749.1
          XM_047424794.1 3 872 XP_047280750.1
          XM_047424795.1 3 872 XP_047280751.1
          XM_047424796.1 3 872 XP_047280752.1
          XM_047424797.1 3 872 XP_047280753.1
          XM_047424798.1 3 872 XP_047280754.1
          XM_047424799.1 3 872 XP_047280755.1
          XM_047424800.1 3 872 XP_047280756.1
          XM_047424801.1 2 788 XP_047280757.1
          XM_047424803.1 3 872 XP_047280759.1
          AK097602.1 3 532 BAC05114.1
          BC031618.1 2 331 AAH31618.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv825252 Chr.10:13492728 - 13549437 on Build GRCh38 Gain BEND7
          esv1007603 Chr.10:13489372 - 13500710 on Build GRCh38 Deletion BEND7

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Gene Ontology Categories:

          Process(es)

          DNA binding
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline