Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01908445_cn
          See other CRIP2 CNV Assays ›
          Gene Symbol
          CRIP2
          Assay Reference Genome
          Location

          Chr.14:105479916 on build GRCh38
          Cytoband
          14q32.33
          Assay ID Hs01908445_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1397

          Gene Name:

          cysteine rich protein 2

          Gene Aliases:

          CRIP, CRP2, ESP1

          Location:

          Chr.14:105472938-105480170 on Build GRCh38

          Assay Gene Location:

          Within Exon 11
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CRIP2 NM_001270837.1 8 1422 NP_001257766.1
          NM_001270841.1 4 615 NP_001257770.1
          NM_001312.3 8 978 NP_001303.1
          NR_073081.1 6 779
          NR_073082.1 8 954
          NR_073083.1 7 883
          NR_073084.1 8 1731
          NR_073085.1 8 1203
          AK057837.1 7 755
          AK091845.1 8 1731
          AK223251.1 8 890 BAD96971.1
          AW173606.1 1 270
          BC000434.2 8 909 AAH00434.1
          BC001931.1 8 909 AAH01931.1
          BC034151.1 8 959 AAH34151.1
          BC128101.1 8 933
          D42123.1 8 916 BAA07703.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv3996n54 Chr.14:105474226 - 105488519 on Build GRCh38 Gain CRIP1 CRIP2
          nsv817647 Chr.14:105382554 - 105480505 on Build GRCh38 Gain TEX22 PACS2 MTA1 CRIP2 LOC100507437
          dgv3998n54 Chr.14:105474495 - 105770768 on Build GRCh38 Loss IGHG2 IGHG3 IGH IGHA1 ELK2AP MIR8071-1 C14orf80 MIR8071-2 IGHG1 IGHG4 IGHA2 CRIP1 TMEM121 LOC107987211 CRIP2 LOC105370697
          nsv1160204 Chr.14:105291411 - 105483783 on Build GRCh38 Duplication TEX22 PACS2 MTA1 CRIP2 BRF1 LOC100507437
          nsv566229 Chr.14:105474172 - 105483783 on Build GRCh38 Loss CRIP2
          nsv566206 Chr.14:105439736 - 105543235 on Build GRCh38 Loss CRIP1 TMEM121 MTA1 CRIP2 C14orf80 LOC105370697
          nsv832890 Chr.14:105304955 - 105483089 on Build GRCh38 Loss TEX22 PACS2 MTA1 CRIP2 BRF1 LOC100507437
          nsv1440 Chr.14:105473170 - 105479862 on Build GRCh38 Insertion CRIP2
          nsv566210 Chr.14:105465248 - 105488368 on Build GRCh38 Gain CRIP1 MTA1 CRIP2
          nsv9177 Chr.14:105476516 - 105485998 on Build GRCh38 Gain CRIP2
          nsv827088 Chr.14:105454469 - 105498549 on Build GRCh38 Gain CRIP1 MTA1 CRIP2 C14orf80
          nsv952282 Chr.14:105363764 - 105510163 on Build GRCh38 Deletion TEX22 PACS2 CRIP1 MTA1 CRIP2 C14orf80 LOC100507437
          dgv3999n54 Chr.14:105474867 - 105487998 on Build GRCh38 Loss CRIP1 CRIP2
          esv2760014 Chr.14:105047009 - 105483089 on Build GRCh38 Loss NUDT14 LOC102723354 TEX22 PACS2 BTBD6 MTA1 JAG2 GPR132 CRIP2 MIR6765 BRF1 LOC100507437
          nsv470679 Chr.14:105124547 - 105605978 on Build GRCh38 Loss NUDT14 IGH TEX22 JAG2 C14orf80 BRF1 LOC100507437 IGHA2 PACS2 BTBD6 CRIP1 TMEM121 MTA1 CRIP2 MIR6765 LOC105370697
          dgv3987n54 Chr.14:105461599 - 105499970 on Build GRCh38 Loss CRIP1 MTA1 CRIP2 C14orf80
          nsv1054190 Chr.14:105246882 - 105701138 on Build GRCh38 Gain IGHG2 IGH TEX22 ELK2AP MIR8071-1 C14orf80 BRF1 LOC100507437 MIR8071-2 IGHG4 IGHA2 PACS2 BTBD6 CRIP1 TMEM121 MTA1 CRIP2 LOC105370697
          esv3635645 Chr.14:105473682 - 105484540 on Build GRCh38 Gain CRIP2
          nsv517055 Chr.14:105136282 - 106779068 on Build GRCh38 Gain+Loss LINC00226 IGH TEX22 IGHV3-48 MIR4507 MIR4537 IGHG1 IGHG4 IGHA2 MIR4539 PACS2 IGHV4-55 CRIP1 TMEM121 ADAM6 CRIP2 LOC105370697 MIR4538 NUDT14 IGHV1-69 IGHG2 IGHG3 IGHA1 ELK2AP IGHV3-23 IGHM JAG2 IGHV3-30 MIR8071-1 IGHD C14orf80 BRF1 LOC100507437 MIR8071-2 KIAA0125 IGHV3-41 IGHV3-71 BTBD6 LINC00221 MTA1 IGHV4-31 LOC107987211 MIR6765 IGHV3-69-1
          dgv3991n54 Chr.14:105472502 - 105480505 on Build GRCh38 Loss CRIP2

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          hemopoiesis
          zinc ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline