Search
Search

Entrez Gene ID: | 641384 |
Gene Name: | long intergenic non-protein coding RNA 2912 |
Gene Aliases: |
TMEM75 |
Location: |
Chr.8:127946559-127948723 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC02912 | NR_103558.1 | 1 | 405 | |
| AK093424.1 | 1 | 405 | ||
| AX748117.1 | 1 | 405 | ||
| BC137383.1 | AAI37384.1 | |||
| BC137385.1 | AAI37386.1 |
Entrez Gene ID: | 5820 |
Gene Name: | Pvt1 oncogene |
Gene Aliases: |
LINC00079, MIR1204HG, NCRNA00079, TP53LC09, onco-lncRNA-100 |
Location: |
Chr.8:127794524-128101256 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5490037 | Chr.8:127933362 - 127951132 on Build GRCh38 | Deletion |
|
| nsv524801 | Chr.8:127943179 - 127953306 on Build GRCh38 | Loss |
|
| nsv3169835 | Chr.8:127935979 - 127950518 on Build GRCh38 | Deletion |
|